Skip to Content
Merck
All Photos(1)

Key Documents

131067

Sigma-Aldrich

4-Hydroxy-1-naphthaldehyde

98%

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
HOC10H7CHO
CAS Number:
Molecular Weight:
172.18
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Assay

98%

mp

179-182 °C (lit.)

functional group

aldehyde

SMILES string

Oc1ccc(C=O)c2ccccc12

InChI

1S/C11H8O2/c12-7-8-5-6-11(13)10-4-2-1-3-9(8)10/h1-7,13H

InChI key

LORPDGZOLAPNHP-UHFFFAOYSA-N

Application

4-Hydroxy-1-naphthaldehyde was used in the preparation of racemic 1- and 2-naphthol analogues of tyrosine, 2-amino-3-(4-hydroxy-1-naphthyl)propanoic acid hydrochloride and 2-amino-3-(6-hydroxy-2-naphthyl)propanoic acid hydrobromide.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Zi-Jian Chen et al.
Ecotoxicology and environmental safety, 196, 110533-110533 (2020-04-05)
1-naphthol (1-NAP) is the main metabolite of pesticide carbaryl and naphthalene, and is also a genotoxic and carcinogenic intermediate in the synthesis of organic compound, dyes, pigment and pharmaceutical industry. In this work, two novel haptens were designed and synthesized
Syntheses of 1-and 2-naphthol analogs of DL-tyrosine. Potential fluorescent probes of peptide structure and dynamics in complex environments.
Vela MA, et al.
The Journal of Organic Chemistry, 55(9), 2913-2918 (1990)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service