Skip to Content
Merck
All Photos(1)

Key Documents

129623

Sigma-Aldrich

2-Mercaptopyrimidine

98%

Synonym(s):

2-Pyrimidinethiol

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C4H4N2S
CAS Number:
Molecular Weight:
112.15
Beilstein:
107105
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Quality Level

Assay

98%

form

powder

mp

230 °C (dec.) (lit.)

SMILES string

Sc1ncccn1

InChI

1S/C4H4N2S/c7-4-5-2-1-3-6-4/h1-3H,(H,5,6,7)

InChI key

HBCQSNAFLVXVAY-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

2-Pyrimidinethiol has been used to study the highly selective palladium catalyzed kinetic resolutions of the racemic cyclic allylic carbonates and racemic acyclic allylic carbonates. It has also been used to study the photodegradation of 2-pyrimidinethiol in aerated aqueous solution using zinc oxide.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Jie Huang et al.
Nanoscale, 2(12), 2733-2738 (2010-10-12)
In this paper, we describe the formation of Au nanoparticle-graphene oxide (Au-GO) and -reduced GO (Au-rGO) composites by noncovalent attachment of Au nanoparticles premodified with 2-mercaptopyridine to GO and rGO sheets, respectively, viaπ-π stacking and other molecular interactions. Compared with
V Sood et al.
Folia morphologica, 37(4), 402-406 (1989-01-01)
The telotrophic ovary of Epilachna vigintioctopunctata is composed of 32-40 ovarioles, each with an apical germarium and a basal vitellarium. The germarium encloses mononucleate and binucleate trophocytes, prefollicular tissue and oogonia, while the vitellarium contains 2-5 oocytes arranged in order
Denis Lima Guerra et al.
Journal of hazardous materials, 166(2-3), 1550-1555 (2009-01-22)
The compound 2-mercaptopyrimidine (MPY) was attached onto synthetic Na-Magadiite (M) and Na-Kanemite (K) samples by homogeneous route. The final matrices named M(MPY) and K(MPY) have been characterized through X-ray powder diffraction and scanning electron microscopy (SEM). The resulted materials were
Morteza Montazerozohori et al.
Annali di chimica, 96(5-6), 285-292 (2006-07-22)
Photodegradation of several thiols and thiones including 2-pyrimidinethiol, thiazolidine-2-thione, imidazolidine-2-thione, 1,2,4- triazole 3-thiol and 2,5-dimercapto- 1,3,4-thiadiazole have been studied in aerated aqueous solution using zinc oxide under irradiation with 400W high pressure mercury lamp. Rate constants span the range from
Barbara Cosimelli et al.
Journal of medicinal chemistry, 54(15), 5597-5601 (2011-07-01)
We report the preliminary in vitro characterization of a series of pyrimidines as a new chemotype that modulates cardiovascular parameters and relaxes ileum smooth muscle according to classical calcium entry blockers. Tested compounds showed an interesting negative inotropic selectivity. In

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service