Skip to Content
Merck
All Photos(1)

Key Documents

124613

Sigma-Aldrich

2-Chloro-4,6-dinitrophenol

97%

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C6H3ClN2O5
CAS Number:
Molecular Weight:
218.55
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Assay

97%

mp

110-114 °C (lit.)

functional group

chloro

SMILES string

Oc1c(Cl)cc(cc1[N+]([O-])=O)[N+]([O-])=O

InChI

1S/C6H3ClN2O5/c7-4-1-3(8(11)12)2-5(6(4)10)9(13)14/h1-2,10H

InChI key

PCBCIXWBAPIVDV-UHFFFAOYSA-N

General description

2-Chloro-4,6-dinitrophenol was added as nitrogen, carbon and energy supplement in the culture medium of Rhodococcus erythropolis HL 24-1.

Application

2-Chloro-4,6-dinitrophenol was used in identification of components in chemical waste water and in effluents from biological wastewater treatment plant using electrospray ionization and tandem mass spectrometry.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service