Skip to Content
Merck
All Photos(4)

Key Documents

121649

Sigma-Aldrich

2-Deoxy-D-ribose

97% (GC)

Synonym(s):

2-Deoxy-D-arabinose, 2-Deoxy-D-erythropentose, Thyminose

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C5H10O4
CAS Number:
Molecular Weight:
134.13
Beilstein:
1721978
EC Number:
MDL number:
UNSPSC Code:
12352201
PubChem Substance ID:
NACRES:
NA.22

Quality Level

Assay

97% (GC)

form

powder

optical activity

[α]22/D −59°, c = 1 in H2O

color

white to off-white

mp

89-90 °C (lit.)

SMILES string

OC[C@@H](O)[C@@H](O)CC=O

InChI

1S/C5H10O4/c6-2-1-4(8)5(9)3-7/h2,4-5,7-9H,1,3H2/t4-,5+/m0/s1

InChI key

ASJSAQIRZKANQN-CRCLSJGQSA-N

Looking for similar products? Visit Product Comparison Guide

Other Notes

To gain a comprehensive understanding of our extensive range of Monosaccharides for your research, we encourage you to visit our Carbohydrates Category page.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Manuel Ellermann et al.
Acta crystallographica. Section D, Biological crystallography, 68(Pt 3), 253-260 (2012-02-22)
The biological activity of catechol neurotransmitters such as dopamine in the synapse is modulated by transporters and enzymes. Catechol-O-methyltransferase (COMT; EC 2.1.1.6) inactivates neurotransmitters by catalyzing the transfer of a methyl group from S-adenosylmethionine to catechols in the presence of
Anita R Peoples et al.
Nucleic acids research, 40(13), 6060-6069 (2012-04-03)
Our mechanistic understanding of damage formation in DNA by the direct effect relies heavily on what is known of free radical intermediates studied by EPR spectroscopy. Bridging this information to stable product formation requires methods with comparable sensitivities, a criterion
Tymofii Yu Nikolaienko et al.
Physical chemistry chemical physics : PCCP, 14(44), 15554-15561 (2012-10-18)
Relaxed force constants (RFC) and vibrational root-mean-square (VRMS) deviations are used for comparative characterization of mechanical properties of canonical 2'-deoxyribonucleosides (2DRs) and 1,2-dideoxyribose molecule, their model sugar residue. It has been shown that RFC and VRMS should be preferred over
Buthina Al-Oudat et al.
Bioorganic & medicinal chemistry letters, 23(3), 854-859 (2012-12-26)
Well-defined substrates for the study of oxidative processes are important for the elucidation of the role of DNA damage in the etiology of diseases such as cancer. We have synthesized 3'-modified oligodeoxyribonucleotides (ODNs) using 5'→3' 'reverse' DNA synthesis for the
Guillaume Mata et al.
The Journal of organic chemistry, 77(20), 9006-9017 (2012-09-15)
An efficient method for the N-2-deoxyribosylation of modified nucleobases by 2-deoxythioriboside donors is reported. In the presence of an in situ silylated nucleobase, thioglycosides can be activated with NIS/HOTf to give nucleosides in high yields and with good β-selectivity. By

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service