Skip to Content
Merck
All Photos(1)

Key Documents

111627

Sigma-Aldrich

2-Methyl-1-hexene

96%

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CH3(CH2)3C(CH3)=CH2
CAS Number:
Molecular Weight:
98.19
Beilstein:
1732738
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

vapor density

1 (vs air)

Quality Level

Assay

96%

refractive index

n20/D 1.403 (lit.)

bp

92 °C (lit.)

density

0.697 g/mL at 25 °C (lit.)

SMILES string

CCCCC(C)=C

InChI

1S/C7H14/c1-4-5-6-7(2)3/h2,4-6H2,1,3H3

InChI key

IRUDSQHLKGNCGF-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

2-Methyl-1-hexene is a less reactive olefin that is hydroformylated in the presence of phosphite-modified rhodium catalysts. The epoxidation of 2-methyl-1-hexene is catalyzed by human and rat P450 enzymes.

Pictograms

FlameHealth hazard

Signal Word

Danger

Hazard Statements

Precautionary Statements

Hazard Classifications

Asp. Tox. 1 - Flam. Liq. 2

Storage Class Code

3 - Flammable liquids

WGK

WGK 3

Flash Point(F)

17.6 °F - closed cup

Flash Point(C)

-8 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Hydroformylation of less reactive olefins with modified rhodium catalysts.
Van Leeuwen PWNM and Roobeek CF.
Journal of Organometallic Chemistry, 258(3), 343-350 (1983)
C Chiappe et al.
Chemical research in toxicology, 11(12), 1487-1493 (1998-12-22)
The epoxidation of 1-hexene (1a) and 2-methyl-1-hexene (1b), two hydrocarbons present in the ambient air as pollutants, is catalyzed by some human and rat P450 enzymes. The enantioselectivities of these processes, when the reactions were carried out using rat and
Hiroshi Toda et al.
Applied and environmental microbiology, 81(6), 1919-1925 (2015-01-04)
We describe the development of biocatalysis for producing optically pure straight-chain (S)-epoxyalkanes using styrene monooxygenase of Rhodococcus sp. strain ST-10 (RhSMO). RhSMO was expressed in the organic solvent-tolerant microorganism Kocuria rhizophila DC2201, and the bioconversion reaction was performed in an

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service