Skip to Content
Merck
All Photos(1)

Key Documents

111279

Sigma-Aldrich

4-Penten-1-ol

99%

Synonym(s):

2-Allylethyl alcohol

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CH2=CH(CH2)3OH
CAS Number:
Molecular Weight:
86.13
Beilstein:
1560163
EC Number:
MDL number:
UNSPSC Code:
12352100
eCl@ss:
39020310
PubChem Substance ID:
NACRES:
NA.22

Assay

99%

refractive index

n20/D 1.429 (lit.)

bp

134-137 °C (lit.)

density

0.834 g/mL at 25 °C (lit.)

functional group

allyl
hydroxyl

SMILES string

OCCCC=C

InChI

1S/C5H10O/c1-2-3-4-5-6/h2,6H,1,3-5H2

InChI key

LQAVWYMTUMSFBE-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

4-penten-1-ol forms ester bond at the C terminus of the linear peptide in solution with HATU as coupling agent.

Application

4-Penten-1-ol can be used as a reactant to prepare sulfamate ester by reacting with chlorosulfonyl isocycanate (142662).The derived ester undergoes an enantioselective intramolecular azridination reaction in the presence of Cu catalyst. 4-Penten-1-ol can also be used to study the epoxidation of olefins with oxo-diperoxo tungstate(VI) complex as catalyst and bicarbonate as co-catalyst.

Pictograms

Flame

Signal Word

Warning

Hazard Statements

Hazard Classifications

Flam. Liq. 3

Storage Class Code

3 - Flammable liquids

WGK

WGK 3

Flash Point(F)

118.4 °F - closed cup

Flash Point(C)

48 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Enantioselective Intramolecular Copper-Catalyzed Aziridination of Sulfamates
Audrey Esteoule.et al.
Synthesis, 1251-1251 (2007)
Highly efficient epoxidation method of olefins with hydrogen peroxide as terminal oxidant, bicarbonate as a co-catalyst and oxodiperoxo molybdenum(VI) complex as catalyst.
Maiti SK, et al.
New. J. Chem., 30(3), 479-489 (2006)
Marina D Rvovic et al.
Journal of molecular modeling, 17(6), 1251-1257 (2010-08-17)
The mechanism of phenylselenoetherification of pent-4-en-1-ol using some bases (pyridine, triethylamine, quinoline, 2,2'-bipyridine) as catalyst was examined through studies of kinetics of the cyclization, by UV-VIS spectrophotometry. It was demonstrated that the intramolecular cyclization is facilitated in the presence of
Stefania Terracciano et al.
Bioorganic & medicinal chemistry, 16(13), 6580-6588 (2008-05-30)
In the recent years, we focused our attention on the cyclodepsipeptide Jaspamide 1, an interesting marine metabolite, possessing a potent inhibitory activity against breast and prostate cancer, as a consequence of its ability to disrupt actin cytoskeleton dynamics. Although its

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service