Skip to Content
Merck
All Photos(1)

Key Documents

110574

Sigma-Aldrich

2,4-Dimethylhexane

99%

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CH3CH2CH(CH3)CH2CH(CH3)2
CAS Number:
Molecular Weight:
114.23
Beilstein:
1718748
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

vapor pressure

48 mmHg ( 34 °C)

Quality Level

Assay

99%

refractive index

n20/D 1.395 (lit.)

bp

108-109 °C (lit.)

density

0.701 g/mL at 25 °C (lit.)

SMILES string

CCC(C)CC(C)C

InChI

1S/C8H18/c1-5-8(4)6-7(2)3/h7-8H,5-6H2,1-4H3

InChI key

HDGQICNBXPAKLR-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

2,4-Dimethylhexane can be gas chromatographically enantioseparated on different modified cyclodextrins.

Signal Word

Danger

Hazard Statements

Hazard Classifications

Aquatic Acute 1 - Aquatic Chronic 1 - Asp. Tox. 1 - Flam. Liq. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Central nervous system

Storage Class Code

3 - Flammable liquids

WGK

WGK 2

Flash Point(F)

71.6 °F - closed cup

Flash Point(C)

22 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Giuseppe Sicoli et al.
Chirality, 21(1), 183-198 (2008-08-14)
The chromatographic enantioseparation of small unfunctionalized chiral alkanes C*HR(1)R(2)R(3) (R = alkyl) represents a challenge in separation science. Because of the lack of any functional groups, enantiorecognition in the presence of a chiral selector is solely based upon weak enantioselective

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service