Skip to Content
Merck
All Photos(1)

Key Documents

110434

Sigma-Aldrich

1-Cyclohexylethanol

97%

Synonym(s):

α-Methylcyclohexanemethanol, (1-Hydroxyethyl)cyclohexane

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CH3CH(C6H11)OH
CAS Number:
Molecular Weight:
128.21
Beilstein:
2322053
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Assay

97%

refractive index

n20/D 1.465 (lit.)

bp

189 °C (lit.)

density

0.928 g/mL at 25 °C (lit.)

functional group

hydroxyl

SMILES string

CC(O)C1CCCCC1

InChI

1S/C8H16O/c1-7(9)8-5-3-2-4-6-8/h7-9H,2-6H2,1H3

InChI key

JMSUNAQVHOHLMX-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

1-Cyclohexylethanol undergoes photocatalytic oxidation (PCO) using nanoparticles titanium dioxide (TiO2-P25) as a semiconductor photocatalyst in acetonitrile, under aerated conditions. It forms cyclohexylethanols by undergoing catalytic ring hydrogenation over several noble metal catalysts supported on charcoal, or on γ-alumina in supercritical carbon dioxide.

Storage Class Code

10 - Combustible liquids

WGK

WGK 2

Flash Point(F)

163.4 °F - closed cup

Flash Point(C)

73 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Slide 1 of 1

1 of 1

Nanoparticles TiO2-photocatalyzed oxidation of selected cyclohexyl alcohols.
Mohamed OS, et al.
Journal of Photochemistry and Photobiology A: Chemistry, 200(2-3), 209-215 (2008)
Sato T, et al.
Journal of Supercritical Fluids, 37(1), 87-93 (2006)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service