Skip to Content
Merck
All Photos(2)

Key Documents

101281

Sigma-Aldrich

3-Bromopropionic acid

97%

Synonym(s):

2-Carboxyethyl bromide, 3-Bromopropanoic acid, beta-bromopropionic acid

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
BrCH2CH2COOH
CAS Number:
Molecular Weight:
152.97
Beilstein:
1071333
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Quality Level

Assay

97%

mp

58-62 °C (lit.)

density

1.48 g/mL at 25 °C (lit.)

SMILES string

OC(=O)CCBr

InChI

1S/C3H5BrO2/c4-2-1-3(5)6/h1-2H2,(H,5,6)

InChI key

DHXNZYCXMFBMHE-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

Quaternization agent in amperometric biosensors. Alkylates mercaptans and other sulfur containing compounds.

Pictograms

Corrosion

Signal Word

Danger

Hazard Statements

Hazard Classifications

Eye Dam. 1 - Skin Corr. 1B

Storage Class Code

8A - Combustible corrosive hazardous materials

WGK

WGK 3

Flash Point(F)

150.8 °F - closed cup

Flash Point(C)

66 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

R L Brown et al.
Biochemistry, 32(38), 10089-10095 (1993-09-28)
Analogs of cGMP bearing diverse substituents at the C8 position of the guanine ring system have been shown to activate the cGMP-activated channel of retinal rods at concentrations lower than cGMP itself. In an effort to understand this behavior, we
Mona Doshi et al.
Molecular pharmacology, 95(5), 475-489 (2019-03-08)
Near-infrared (NIR) imaging is a promising technique for use as a noninvasive and sensitive diagnostic tool. Although the NIR fluorescently labeled glucose analog glucosamine (cypate-glucosamine) has applications in preclinical imaging, the transport pathways and fate of this probe in tissues
B A Gregg et al.
Analytical chemistry, 62(3), 258-263 (1990-02-01)
Oxidoreductases, such as glucose oxidase, can be electrically "wired" to electrodes by electrostatic complexing or by covalent binding of redox polymers so that the electrons flow from the enzyme, through the polymer, to the electrode. We describe two materials for
Synthetic Communications, 23, 373-373 (1993)
Patrick Giraudeau et al.
Journal of magnetic resonance (San Diego, Calif. : 1997), 190(2), 339-345 (2007-12-18)
Recent ultrafast techniques enable nD NMR spectra to be obtained in a single scan. However, resolution enhancement in the ultrafast domain leads to important sensitivity losses and lineshape distortions. In order to understand better resolution and spatial encoding aspects of

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service