Soil, Solid Waste & Groundwater Testing
Soil and groundwater testing are crucial to monitor industrial discharges that may adversely affect the environment and human health. There are numerous technical articles, protocols, and approved analytical methods available to guide solid waste management and testing for heavy metals and other environmental pollutants.
Featured Categories
Explore Spectroquant® solutions: instruments, software, test kits, accessories. From start to finish, ensure rapid, accurate results with user-friendly handling.
Achieve precise separations with our extensive HPLC column collection. Enhance retention, resolution, and selectivity. Order today.
Achieve superior GC performance with our columns, consumables, and accessories. Explore Supelco® offerings.
Certified Reference Materials: CRMs for forensic, clinical, environmental, pharmaceutical, and food & beverage testing.
Solid waste and its impact on soil and groundwater
Solid waste is the refuse generated from human activities during industrial, commercial, mining, and agricultural operations. With an increase in population and industrial activities, solid waste must be appropriately managed and contained to avoid an adverse impact on the environment and human health.
Traditionally, landfill sites are used for solid waste management because of their simplicity, large handling capacity, and minimal operating costs. However, improperly maintained landfills and dumping sites with poor leachate collection systems become a potential source of soil, surface and groundwater contamination. The composition of solid waste can alter soil chemistry and cause a considerable environmental impact by producing leachate and biogas. The most common contaminants found in solid waste leachate are chromium, dioxin, hydrocarbons, organochlorines, PAH, PCB, pesticides, radionuclides, TPH, VOC, persistent organic pollutants (POPs) as well as deadly pathogens. Hence, employing a proper leachate monitoring program is obligatory for the safety and risk assessment.
Soil and Groundwater Testing
Soil and groundwater testing are a hallmark of good leachate monitoring programs and critical in site investigations or environmental risk assessments. Soil and groundwater matrices and chemistries are complex and highly variable, making accurate measurements challenging. The United States Environmental Protection Agency (USEPA) recommends Toxicity Characteristic Leaching Procedure (TCLP) to determine hazardous elements present in waste. This chemical analysis process simulates contaminant leaching in a landfill environment over time, prior to depositing the waste in designated landfills. TCLP determines the mobility of organic and inorganic contaminants (metals, pesticides, herbicides, and solvents) in liquid, solid, and multiphasic wastes.
The Leaching Environmental Assessment Framework (LEAF), is an alternative evaluation system designed to identify and accurately describe the release of inorganic, semi-volatile organic, and non-volatile organic constituents of potential concern (COPCs) in solid materials. LEAF methods were designed to provide a flexible and customizable framework for distinguishing leaching characteristics under a range of conditions for a variety of solid materials in contact with ground or surface water.
Soil and Groundwater Testing Techniques
To ascertain the extent of pollution, regulatory agencies (e.g., USEPA) require the use of official methods when testing soil and groundwater. Soil and groundwater are tested for physicochemical properties, including total dissolved solids, pH, hardness of water, cations, anions, organic matter, total carbon, nitrate, ammonium, and heavy metals. Common analytical techniques for soil and groundwater testing include conductivity, titrimetry, gravimetry, fluorimetry, atomic absorption spectroscopy (AAS), high-performance liquid chromatography (HPLC), gas chromatography, ICP-MS, LC-MS, and GC-MS.
Visit our document search for data sheets, certificates and technical documentation.
Related Articles
- Spectroquant® photometric test accurately quantifies low concentrations of PO4-P in natural waters.
- Neonicotinoids have established themselves as key components in insecticides because of their unique selectivity. The mode of action is similar to nicotine.
- Optimize measurement reliability with advanced analytical quality assurance and process control using Spectroquant®, MQuant®, TraceCERT®, and more.
- This application note describes the fast and efficient separation of the EU’s list of 15 + 1 polynuclear aromatic hydrocarbons (PAHs) using Ascentis® Express PAH HPLC column.
- EMSURE® Grade Inorganic Solvents and Reagents offer exceptional purity and quality control, ensuring reliable, compliant results for regulated analyses.
- See All (18)
Related Protocols
- Ammonium is the naturally preferred source of nitrogen for plant growth and there is an increasing global demand for organic food products.
- Photometric determination with pyridylazoresorcinol (PAR) subsequent to acid extraction
- Photometric determination of ammonium ions in drinking and surface water samples by indophenol blue method using Spectroquant® test kits and spectrophotometer.
- Photometric measurement of ammonium ions in compost samples by indophenol blue method using Spectroquant® test kits and spectrophotometer.
- See All (24)
How Can We Help
In case of any questions, please submit a customer support request
or talk to our customer service team:
Email [email protected]
or call +1 (800) 244-1173
Additional Support
- Chromatogram Search
Use the Chromatogram Search to identify unknown compounds in your sample.
- Calculators & Apps
Web Toolbox - science research tools and resources for analytical chemistry, life science, chemical synthesis and materials science.
- Customer Support Request
Customer support including help with orders, products, accounts, and website technical issues.
- FAQ