Merck
  • Evolution of novel sorbents for effective clean-up of honeybee matrix in highly toxic insecticide LC/MS/MS analysis.
Evolution of novel sorbents for effective clean-up of honeybee matrix in highly toxic insecticide LC/MS/MS analysis.
Ecotoxicology and environmental safety (2017-01-28)
Piotr Kaczynski, Izabela Hrynko, Bozena Lozowicka
ABSTRACT
Highly toxic insecticides (HTIs) belonging to different chemical groups are dangerous to pollinating organisms, even in sublethal doses. An important objective of this study was to develop a method to determine over fifty HTIs at very low concentrations in the bee matrix. The novelty of this research involved obtaining extract completely free from beewaxes, lipids and proteins using EMR-lipid (enhanced matrix removal-lipid), chitin and Z-Sep+ (zirconium oxide and C18 dual-bonded to silica) as clean-up sorbents. Different parameters, such as weight of bees, extraction solvent, and freezing time were evaluated. Determinations were made using liquid chromatography-tandem mass spectrometry (LC-MS/MS). EMR-lipid allows for removing most of the fatty co-extracts and increases the overall performance of the method by reducing the matrix effects (ME) without significant analyte loss. The established modified QuEChERS method based on 1% acetic acid in acetonitrile extraction followed by EMR-lipid clean-up was validated at three different spiking levels (0.001, 0.01 and 0.1mgkg
Product Description
Supelco
Supel QuE, Z-Sep+ Bulk, pkg of 20 g
Supelco
Supel QuE, Z-Sep/C18 Tube, centrifuge tube volume 2 mL, pack of 100
Supelco
Supel QuE, Z-Sep+ Tube, centrifuge tube volume 2 mL, pack of 100 ea
Supelco
Supel QuE, Z-Sep/C18 Tube, pk of 50, centrifuge tube volume 15 mL , Shaker Compatible
Supelco
Supel QuE, Z-Sep+ with MgSO4 Tube, pk of 50, centrifuge tube volume 15 mL , Shaker Compatible
Supelco
Supel QuE, Z-Sep Tube, pk of 50, centrifuge tube volume 15 mL , Shaker Compatible
Supelco
Supel QuE, Z-Sep Bulk
Supelco
Supel QuE, Z-Sep Tube, centrifuge tube volume 2 mL, pack of 100 ea
Supelco
Supel QuE, Z-Sep+ Tube, pk of 50, centrifuge tube volume 15 mL , Shaker Compatible