Skip to Content
Merck
All Photos(1)

Documents

55299-U

Supelco

Supel QuE QuEChERS tube

Z-Sep+ Bulk, pkg of 20 g

Synonym(s):

QuEChERS

Sign Into View Organizational & Contract Pricing


About This Item

EC Number:
UNSPSC Code:
41115712
NACRES:
NB.21

product name

Supel QuE, Z-Sep+ Bulk, pkg of 20 g

material

polypropylene tube

Quality Level

product line

Supel

composition

Z-Sep+ Bulk

packaging

pkg of 20 g

technique(s)

QuEChERS: suitable

surface area

480 m2/g

matrix

silica gel base material

matrix active group

zirconia-based phase

particle size

50 μm

pore size

70 Å pore size

application(s)

food and beverages

Looking for similar products? Visit Product Comparison Guide

General description

Dispersive SPE (dSPE), often referred to as the "QuEChERS" method (Quick, Easy, Cheap, Effective, Rugged, and Safe), is modern sample prep technique that is becoming increasingly popular in the area of multi-residue pesticide analysis in food and agricultural products.

Using the QuEChERS method, food/agricultural samples are first extracted with an aqueous miscible solvent (e.g., acetonitrile) in the presence of high amounts of salts (e.g., sodium chloride and magnesium sulfate) and/or buffering agents (e.g. citrate) to induce liquid phase separation and stabilize acid and base labile pesticides, respectively. Upon shaking and centrifugation, an aliquot of the organic phase is subjected to further cleanup using SPE. Unlike traditional methods using SPE tubes, in dispersive SPE, cleanup is facilitated by mixing bulk amounts of SPE (e.g., Supelclean PSA, ENVI-Carb, and/or Discovery DSC-18) with the extract. After sample cleanup, the mixture is centrifuged and the resulting supernatant can either be analyzed directly or can be subjected to minor further treatment before analysis.

Supelco carries a line of vials and centrifuge tubes containing pre-determined amounts of salts and SPE sorbents to support the most common method configurations used today.

Other Notes

SupelQuE Z-Sep+ enhances sample cleanup for complex matrices by effectively removing more fat and color from sample extracts than traditional phases for QuEChERS LC-MS methods. By eliminating problematic matrix interferences, Z-Sep products provide more robust LC-MS methods. This proprietary technology can replace C18 and PSA in your current methods without additional method development.

Z-Sep+ is recommended for cleanup of samples containing greater than 15% fat.

Legal Information

Certain uses of this product may be covered by the claims of one or more pending patent applications. For licensing inquiries please contact techserv@sial.com
Supel is a trademark of Sigma-Aldrich Co. LLC

Storage Class Code

11 - Combustible Solids

WGK

nwg

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Keiko Iwakoshi et al.
Shokuhin eiseigaku zasshi. Journal of the Food Hygienic Society of Japan, 57(5), 150-154 (2016-10-28)
Rapid multi-residue analysis of pesticides in pulses was developed using LC-MS/MS. Pesticide residues in 5 g of homogenized pulses were extracted with 30 mL of acetonitrile and salted out with 4 g of anhydrous magnesium sulfate and 2 g of
Pol Herrero et al.
Journal of chromatography. A, 1339, 34-41 (2014-03-25)
A Quick, Easy, Cheap, Effective, Rugged and Safe (QuEChERS) extraction method followed by liquid chromatography-(Orbitrap) high resolution mass spectrometry was developed for the simultaneous determination of five benzotriazole, four benzothiazole and five benzenesulfonamide derivates in sewage sludge. While the method
Łukasz Rajski et al.
Journal of chromatography. A, 1304, 109-120 (2013-07-23)
Several extraction methods were evaluated in terms of recoveries and extraction precision for 113 pesticides in avocado: QuEChERS with various d-SPE clean-ups (Z-Sep, Z-Sep+, PSA+C18 and silica), miniLuke and ethyl acetate. Extracts were analysed using liquid chromatography coupled with triple
Tomasz Tuzimski et al.
Journal of AOAC International, 100(6), 1671-1680 (2017-07-15)
In this study, a simple, cost-effective, and sensitive HPLC diode-array detection method was developed for the simultaneous determination of six different 5-nitroimidazoles [metronidazole, 2-hydroxymethyl-1-methyl-5-nitro-1H-imidazole, dimetridazole (DMZ), ronidazole, ornidazole, and ipronidazole] in bovine milk samples. A QuEChERS-based sample preparation procedure was
Hui Li et al.
Journal of chromatography. A, 1528, 53-60 (2017-11-07)
Herein, an accurate and reliable isotope-labelled internal standard method was developed and validated for simultaneous determination of three polar pesticide adjuvants, namely 2-pyrrolidone, N-methyl-2-pyrrolidone, and N-ethyl-2-pyrrolidone in plant-derived agro-products. Matrices, including apple, cabbage, tomato, cucumber, rice, and wheat were extracted

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service