Merck
  • GORAB Missense Mutations Disrupt RAB6 and ARF5 Binding and Golgi Targeting.
GORAB Missense Mutations Disrupt RAB6 and ARF5 Binding and Golgi Targeting.
The Journal of investigative dermatology (2015-05-23)
Johannes Egerer, Denise Emmerich, Björn Fischer-Zirnsak, Wing Lee Chan, David Meierhofer, Beyhan Tuysuz, Katrin Marschner, Sascha Sauer, Francis A Barr, Stefan Mundlos, Uwe Kornak
ABSTRACT
Gerodermia osteodysplastica is a hereditary segmental progeroid disorder affecting skin, connective tissues, and bone that is caused by loss-of-function mutations in GORAB. The golgin, RAB6-interacting (GORAB) protein localizes to the Golgi apparatus and interacts with the small GTPase RAB6. In this study, we used different approaches to shed more light on the recruitment of GORAB to this compartment. We show that GORAB best colocalizes with trans-Golgi markers and is rapidly displaced upon Brefeldin A exposition, indicating a loose association with Golgi membranes. A yeast two-hybrid screening revealed a specific interaction with the small GTPase ADP-ribosylation factor (ARF5) in its active, GTP-bound form. ARF5 and RAB6 bind to GORAB via the same internal Golgi-targeting RAB6 and ARF5 binding (IGRAB) domain. Two GORAB missense mutations identified in gerodermia osteodysplastica patients fall within this IGRAB domain. GORAB carrying the mutation p.Ala220Pro had a cytoplasmic distribution and failed to interact with both RAB6 and ARF5. In contrast, the p.Ser175Phe mutation displaced GORAB from the Golgi compartment to vesicular structures and selectively impaired ARF5 binding. Our findings indicate that the IGRAB domain is crucial for the Golgi localization of GORAB and that loss of this localization impairs its physiological function.
Sigma-Aldrich
Ethylenediaminetetraacetic acid, SAJ special grade, ≥99.0%
Sigma-Aldrich
Sodium hydroxide-16O solution, 20 wt. % in H216O, 99.9 atom % 16O
Sigma-Aldrich
Ethylenediaminetetraacetic acid, anhydrous, crystalline, BioReagent, suitable for cell culture
Sigma-Aldrich
Ethylenediaminetetraacetic acid, purified grade, ≥98.5%, powder
Sigma-Aldrich
Magnesium chloride, anhydrous, ≥98%
Sigma-Aldrich
Magnesium chloride, BioReagent, suitable for insect cell culture, ≥97.0%
Sigma-Aldrich
Ethylenediaminetetraacetic acid, ACS reagent, 99.4-100.6%, powder
Sigma-Aldrich
L-Glutathione reduced, ≥98.0%
Sigma-Aldrich
Ethylenediaminetetraacetic acid solution, 0.02% in DPBS (0.5 mM), sterile-filtered, BioReagent, suitable for cell culture
Sigma-Aldrich
Magnesium chloride, AnhydroBeads, −10 mesh, 99.99% trace metals basis
Sigma-Aldrich
Magnesium chloride, powder, <200 μm
Sigma-Aldrich
Ethylenediaminetetraacetic acid, 99.995% trace metals basis
Sigma-Aldrich
Magnesium chloride, AnhydroBeads, −10 mesh, 99.9% trace metals basis
Sigma-Aldrich
L-Glutathione reduced, BioXtra, ≥98.0%
Sigma-Aldrich
L-Glutathione reduced, suitable for cell culture, BioReagent, ≥98.0%, powder
Sigma-Aldrich
Ethylenediaminetetraacetic acid, BioUltra, anhydrous, ≥99% (titration)
Sigma-Aldrich
Anti-GORAB antibody produced in rabbit, Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution, ab1
Sigma-Aldrich
Sodium chloride solution, 0.85%
Sigma-Aldrich
Sodium chloride, SAJ first grade, ≥99.0%
Sigma-Aldrich
Sodium hydroxide solution, 1 M
Sigma-Aldrich
Sodium hydroxide solution, 0.01 M
Sigma-Aldrich
Sodium hydroxide solution, 0.05 M
Sigma-Aldrich
Sodium hydroxide solution, 0.1 M
Sigma-Aldrich
Sodium hydroxide solution, 4 M
Sigma-Aldrich
Sodium chloride, JIS special grade, ≥99.5%
Sigma-Aldrich
Sodium hydroxide solution, 6 M
Sigma-Aldrich
Sodium chloride solution, 1 M
Sigma-Aldrich
Sodium chloride solution, 0.1 M
Sigma-Aldrich
Sodium hydroxide solution, 7 M
Sigma-Aldrich
Sodium chloride, random crystals, optical grade, 99.9% trace metals basis