Merck
  • Trastuzumab emtansine (T-DM1) renders HER2+ breast cancer highly susceptible to CTLA-4/PD-1 blockade.
Trastuzumab emtansine (T-DM1) renders HER2+ breast cancer highly susceptible to CTLA-4/PD-1 blockade.
Science translational medicine (2015-11-27)
Philipp Müller, Matthias Kreuzaler, Tarik Khan, Daniela S Thommen, Kea Martin, Katharina Glatz, Spasenija Savic, Nadia Harbeck, Ulrike Nitz, Oleg Gluz, Michael von Bergwelt-Baildon, Hans Kreipe, Sai Reddy, Matthias Christgen, Alfred Zippelius
ABSTRACT
Targeted drug delivery with antibody-drug conjugates such as the HER2-directed ado-trastuzumab emtansine (T-DM1) has emerged as a powerful strategy for cancer therapy. We show that T-DM1 is particularly effective in eliciting antitumor immunity in patients with early breast cancer (WSG-ADAPT trial) and in a HER2-expressing orthotopic tumor model. In the latter, despite primary resistance to immunotherapy, combined treatment with T-DM1 and anti-CTLA-4/PD-1 (cytotoxic T lymphocyte-associated protein-4/programmed cell death protein-1) was curative because it triggered innate and adaptive immunity. Tumor rejection was accompanied by massive T cell infiltration, TH1 (T helper 1) cell polarization, and, notably, a substantial increase in regulatory T cells. Depletion of regulatory T cells resulted in inflammation and tissue damage, implying their essential role in protecting the host during therapy. This study provides insights into the mechanisms of T-DM1's therapeutic activity and a rationale for potential therapeutic combination strategies with immunotherapy.
Sigma-Aldrich
2-Mercaptoethanol, ≥99.0%
Sigma-Aldrich
2-Mercaptoethanol, SAJ special grade, ≥99.0%
Sigma-Aldrich
Phorbol 12-myristate 13-acetate, synthetic, ≥98.0% (TLC)
Sigma-Aldrich
2-Mercaptoethanol, BioUltra, for molecular biology, ≥99.0% (GC)
Sigma-Aldrich
Sodium pyruvate, BioXtra, ≥99%
Sigma-Aldrich
Sodium pyruvate, powder, BioReagent, suitable for cell culture, suitable for insect cell culture, ≥99%
Sigma-Aldrich
2-Mercaptoethanol, ≥99.0%
Sigma-Aldrich
Thymine 1-β-D-arabinofuranoside, ≥99%
Sigma-Aldrich
PMA, for use in molecular biology applications, ≥99% (HPLC)
Sigma-Aldrich
2-Mercaptoethanol, for molecular biology, suitable for electrophoresis, suitable for cell culture, BioReagent, 99% (GC/titration)
Sigma-Aldrich
Sodium pyruvate, powder, BioXtra, suitable for mouse embryo cell culture
Sigma-Aldrich
Sodium pyruvate, Hybri-Max, powder, suitable for hybridoma
Sigma-Aldrich
Sodium pyruvate, SAJ special grade, ≥95.0%
Sigma-Aldrich
Sodium pyruvate, ReagentPlus®, ≥99%
Sigma-Aldrich
Phorbol 12-myristate 13-acetate, ≥99% (TLC), film or powder
Sigma-Aldrich
Sodium pyruvate, anhydrous, free-flowing, Redi-Dri, ReagentPlus®, ≥99%
Sigma-Aldrich
L-Glutamine
Sigma-Aldrich
Phenylacetic acid, 99%
Sigma-Aldrich
Phenylacetic acid, ≥99%, FCC, FG
Sigma-Aldrich
L-Glutamine, BioUltra, ≥99.5% (NT)
SAFC
L-Glutamine
Sigma-Aldrich
L-Glutamine, γ-irradiated, BioXtra, suitable for cell culture
Sigma-Aldrich
L-Glutamine, meets USP testing specifications, suitable for cell culture, 99.0-101.0%, from non-animal source
Sigma-Aldrich
L-Glutamine