Merck
  • Low DICER1 expression is associated with attenuated neutrophil differentiation and autophagy of NB4 APL cells.
Low DICER1 expression is associated with attenuated neutrophil differentiation and autophagy of NB4 APL cells.
Journal of leukocyte biology (2015-05-21)
Julian Wampfler, Elena A Federzoni, Bruce E Torbett, Martin F Fey, Mario P Tschan
ABSTRACT
Successful myeloid differentiation depends on the expression of a series of miRNAs. Thus, it is hardly surprising that miRNAs are globally repressed in AML, a disease mainly characterized by a block in cellular myeloid differentiation. Studies investigating the mechanisms for low miRNA expression in AML has mostly focused on altered transcriptional regulation or deletions, whereas defective miRNA processing has received less attention. In this study, we report that the expression of the key miRNA processing enzyme DICER1 is down-regulated in primary AML patient samples and healthy CD34(+) progenitor cells as compared with granulocytes. In line with these findings, Dicer1 expression was induced significantly in AML cell lines upon neutrophil differentiation. The knocking down of DICER1 in AML cells significantly attenuated neutrophil differentiation, which was paralleled by decreased expression of miRNAs involved in this process. Moreover, we found that inhibiting DICER1 attenuated the activation of autophagy, a cellular recycling process that is needed for proper neutrophil differentiation of AML cells. Our results clearly indicate that DICER1 plays a novel role in neutrophil differentiation as well as in myeloid autophagy of AML cells.
Sigma-Aldrich
Urea-12C, 99.9 atom % 12C
Sigma-Aldrich
Penicillin-Streptomycin, Solution stabilized, with 10,000 units penicillin and 10 mg streptomycin/mL, 0.1 μm filtered, BioReagent, suitable for cell culture
Sigma-Aldrich
Urea solution, 40 % (w/v) in H2O
Sigma-Aldrich
Urea solution, BioUltra, ~8 M in H2O
Sigma-Aldrich
Urea, powder, BioReagent, for molecular biology, suitable for cell culture
Sigma-Aldrich
Urea, suitable for electrophoresis
Sigma-Aldrich
Urea, BioXtra, pH 7.5-9.5 (20 °C, 5 M in H2O)
Sigma-Aldrich
Urea, meets USP testing specifications
Sigma-Aldrich
Urea, ACS reagent, 99.0-100.5%
Sigma-Aldrich
Urea, ReagentPlus®, ≥99.5%, pellets
Sigma-Aldrich
Urea, BioUltra, for molecular biology, 99% (T)
Sigma-Aldrich
Urea, puriss. p.a., ACS reagent, reag. Ph. Eur., ≥99%
Sigma-Aldrich
Urea, puriss., meets analytical specification of Ph. Eur., BP, USP, 99.0-100.5%, 99.0-101.0% (calc. on dry substance)
Sigma-Aldrich
Urea, ≥99.0%
Sigma-Aldrich
MISSION® pLKO.1-puro Non-Mammalian shRNA Control Plasmid DNA, Targets no known mammalian genes
Sigma-Aldrich
Urea, SAJ first grade, ≥98.0%
Sigma-Aldrich
Urea, JIS special grade, ≥99.0%
Supelco
Urea, 8 M (after reconstitution with 16 mL high purity water)
Sigma-Aldrich
Anti-Glyceraldehyde-3-Phosphate Dehydrogenase Antibody, clone 6C5, clone 6C5, Chemicon®, from mouse