Merck
  • Combined negative effect of donor age and time in culture on the reprogramming efficiency into induced pluripotent stem cells.
Combined negative effect of donor age and time in culture on the reprogramming efficiency into induced pluripotent stem cells.
Stem cell research (2015-06-23)
Ras Trokovic, Jere Weltner, Parinya Noisa, Taneli Raivio, Timo Otonkoski
ABSTRACT
Somatic cells can be reprogrammed into induced pluripotent stem cells (iPSC) by the forced expression of the transcription factors OCT4, SOX2, KLF4 and c-MYC. Pluripotent reprogramming appears as a slow and inefficient process because of genetic and epigenetic barriers of somatic cells. In this report, we have extended previous observations concerning donor age and passage number of human fibroblasts as critical determinants of the efficiency of iPSC induction. Human fibroblasts from 11 different donors of variable age were reprogrammed by ectopic expression of reprogramming factors. Although all fibroblasts gave rise to iPSC colonies, the reprogramming efficiency correlated negatively and declined rapidly with increasing donor age. In addition, the late passage fibroblasts gave less reprogrammed colonies than the early passage cell counterparts, a finding associated with the cellular senescence-induced upregulation of p21. Knockdown of p21 restored iPSC generation even in long-term passaged fibroblasts of an old donor, highlighting the central role of the p53/p21 pathway in cellular senescence induced by both donor age and culture time.
Sigma-Aldrich
DAPI, for nucleic acid staining
Pricing and availability is not currently available.
Sigma-Aldrich
Sodium butyrate, ≥98.5% (GC)
Pricing and availability is not currently available.
Sigma-Aldrich
Sodium butyrate, 98%
Pricing and availability is not currently available.
Sigma-Aldrich
Sodium butyrate, Vetec, reagent grade, 99%
Pricing and availability is not currently available.
Sigma-Aldrich
Nitrotetrazolium Blue chloride, powder, electrophoresis grade
Pricing and availability is not currently available.
Sigma-Aldrich
Nitrotetrazolium Blue chloride, ≥90.0% (HPLC)
Pricing and availability is not currently available.