Merck
  • Comprehensive proteome analysis of fresh frozen and optimal cutting temperature (OCT) embedded primary non-small cell lung carcinoma by LC-MS/MS.
Comprehensive proteome analysis of fresh frozen and optimal cutting temperature (OCT) embedded primary non-small cell lung carcinoma by LC-MS/MS.
Methods (San Diego, Calif.) (2015-02-28)
Wen Zhang, Shingo Sakashita, Paul Taylor, Ming S Tsao, Michael F Moran
ABSTRACT
Clinical tissue samples provide valuable information for understanding human diseases. One major type of clinical tissue sample that is amenable to various kinds of analysis is fresh frozen and optimal cutting temperature (OCT)-embedded primary patient tissue. Recent advances in mass spectrometry (MS) technologies have been widely applied to study human proteomes by using clinical specimens. However, polymeric compounds such as OCT can interfere with MS analyses. Here we present methods that enable the preparation and analysis of fresh frozen and OCT embedded primary tissue samples by LC-MS/MS. A scraping method was first introduced to reduce the heterogeneity of OCT-embedded non-small cell lung carcinoma tumor sections. OCT compound was reproducibly removed by a series of washing steps involving ethanol and water prior to trypsin digestion. In data-dependent acquisition mode, optimized dynamic exclusion duration settings were established to maximize peptide identifications. These sample preparation conditions and MS parameter settings should be utilized or carefully adjusted in order to achieve optimal comprehensive proteome characterization starting from fresh frozen and OCT embedded clinical tissue specimens.
Sigma-Aldrich
Urea solution, 40 % (w/v) in H2O
Sigma-Aldrich
Bicinchoninic acid disodium salt hydrate, ≥98% (HPLC)
Sigma-Aldrich
Urea solution, BioUltra, ~8 M in H2O
Sigma-Aldrich
Urea-12C, 99.9 atom % 12C
Sigma-Aldrich
Urea, ≥99.0%
Sigma-Aldrich
Urea, SAJ first grade, ≥98.0%
Sigma-Aldrich
Urea, BioUltra, for molecular biology, 99% (T)
Sigma-Aldrich
Urea, puriss. p.a., ACS reagent, reag. Ph. Eur., ≥99%
Sigma-Aldrich
Urea, puriss., meets analytical specification of Ph. Eur., BP, USP, 99.0-100.5%, 99.0-101.0% (calc. on dry substance)
Sigma-Aldrich
Urea, ReagentPlus®, ≥99.5%, pellets
Sigma-Aldrich
Urea, meets USP testing specifications
Sigma-Aldrich
Urea, powder, BioReagent, for molecular biology, suitable for cell culture
Sigma-Aldrich
Urea, BioXtra, pH 7.5-9.5 (20 °C, 5 M in H2O)
Sigma-Aldrich
Urea, suitable for electrophoresis
Sigma-Aldrich
Urea, ACS reagent, 99.0-100.5%
Sigma-Aldrich
Urea, JIS special grade, ≥99.0%
Sigma-Aldrich
Trifluoroacetic acid, SAJ special grade, ≥99.0%
Supelco
Urea, 8 M (after reconstitution with 16 mL high purity water)