Merck
Multilayered thin films from poly(amido amine)s and DNA.
Acta biomaterialia (2015-05-02)
Sry D Hujaya, Johan F J Engbersen, Jos M J Paulusse
ABSTRACT
Dip-coated multilayered thin films of poly(amido amine)s (PAAs) and DNA have been developed to provide surfaces with cell-transfecting capabilities. Three types of PAAs, differing in side chain functional groups, were synthesized and characterized for their properties in forming multilayered structures with ultrasonicated calf thymus DNA (CTDNA) as model DNA. All three polymers display a multilayer build-up in linear profiles as demonstrated by UV spectroscopy. More highly charged side chains were found to provide the lowest deposition of DNA. Surface profiles of the obtained films were investigated by atomic force microscopy (AFM) and static water contact angle measurements to reveal complete surface coverage after at least four layer pair depositions, where alternating patterns of surface profiles were observed depending on whether the cationic polymer or the anionic DNA layer was on top. The stability of the formed surfaces was investigated in vitro under physiological and reductive conditions. Owing to the presence of disulfide bonds in the PAA main chain, the films were readily degraded in the presence of 1mM of DTT in vitro. Under non-reductive physiological conditions, two of the thicker films underwent thermodynamic rearrangement, which resulted in release of approximately half of the incorporated material within 1h, which was caused by the physiological salt concentration. Further, this unpacking phenomenon proved useful in transfecting COS-7 cells seeded on top of these multilayers containing functional plasmid DNA encoding for green fluorescence protein (GFP). Two out of the three different multilayers facilitated good COS-7 cell attachment, proliferation, and transfection in vitro within 2d ays of culture. Fluorescence staining further revealed the presence of DNA-containing released film material among cultured cells. The present work demonstrates the possibility of coating surfaces with thin films that are conveniently adjustable in thickness and amount of active agent to provide cell-transfecting functionality. In this manner transfection can be achieved by simply culturing cells on a multilayer-coated surface in their optimal culture condition (in the presence of serum) and without the need of removing the transfection agent to avoid cytotoxicity.
Sigma-Aldrich
L-(−)-Glucose, ≥99%
Sigma-Aldrich
DL-Dithiothreitol solution, BioUltra, for molecular biology, ~1 M in H2O
Sigma-Aldrich
Hydrogen peroxide solution, 34.5-36.5%
Sigma-Aldrich
Hydrogen peroxide solution, contains potassium stannate as inhibitor, 30-32 wt. % in water, semiconductor grade, 99.999% trace metals basis
Sigma-Aldrich
Sulfuric acid, 99.999%
Sigma-Aldrich
Hydrogen peroxide solution, 30 % (w/w) in H2O, contains stabilizer
Sigma-Aldrich
Sodium phosphate monobasic monohydrate, BioReagent, suitable for electrophoresis, 98.0-102.0%
Sigma-Aldrich
Sulfuric acid solution, 0.05 M
Sigma-Aldrich
di-Sodium hydrogen phosphate dihydrate, BioUltra, for molecular biology, ≥99.0% (T)
Sigma-Aldrich
Sodium phosphate monobasic monohydrate, BioXtra, for molecular biology, ≥99.5% (T)
Sigma-Aldrich
Sulfuric acid, SAJ first grade, ≥95.0%
Sigma-Aldrich
Sulfuric acid solution, 5 mM
Sigma-Aldrich
Sulfuric acid solution, 0.25 M
Sigma-Aldrich
Sulfuric acid solution, 0.025 M
Sigma-Aldrich
Sulfuric acid solution, 0.1 M
Sigma-Aldrich
Sulfuric acid solution, 0.5 M
Sigma-Aldrich
Hydrogen peroxide solution, SAJ first grade, ≥30.0%
Sigma-Aldrich
Sulfuric acid solution, 1.5 M
Sigma-Aldrich
Sulfuric acid, JIS special grade, ≥95.0%
Sigma-Aldrich
Sulfuric acid solution, 0.01 M
Supelco
DL-Dithiothreitol solution, 1 M in H2O