Merck
A SERS-Assisted 3D Barcode Chip for High-Throughput Biosensing.
Small (Weinheim an der Bergstrasse, Germany) (2015-02-18)
Lei Wu, Zhuyuan Wang, Kequan Fan, Shenfei Zong, Yiping Cui
ABSTRACT
A surface enhanced Raman scattering (SERS)-assisted 3D barcode chip has been developed for high-throughput biosensing. The 3D barcode is realized through joint 2D spatial encoding with the Raman spectroscopic encoding, which stores the SERS fingerprint information in the format of a 2D array. Here, the concept of SERS-assisted 3D barcode is demonstrated through multiplex immunoassay, where simultaneous detection of multiple targets in different samples has been achieved using a microfluidic platform. First, multiple proteins in different samples are spatially separated using a microfluidic patterned antibody barcode substrate, forming a 2D hybridization array. Then the SERS probes are used to identify and quantify the proteins. As different SERS probes are labeled with different Raman reporters, they could be employed as "SERS tags" to incorporate spectroscopic information into the 3D barcode. In this 3D barcode, the 2D spatial information helps to differentiate the samples and targets while the SERS information allows quantitative multiplex detection. It is found that the SERS-assisted 3D barcode chip can not only accomplish one-step multiplex detection within 30 min but also achieve an ultrasensitivity down to 10 fg mL(-1) (≈70 aM), which is expected to provide a promising tool for high-throughput biomedical applications.
Sigma-Aldrich
Ethanol, JIS 300, ≥99.5%, suitable for residue analysis
Sigma-Aldrich
Ethanol, ≥99.5%
Sigma-Aldrich
Ethanol, ≥99.5%, suitable for absorption spectrum analysis
Sigma-Aldrich
Ethanol, ≥99.5%, suitable for HPLC
Sigma-Aldrich
Ethanol, ≥99.5%, suitable for fluorescence
Sigma-Aldrich
Ethyl alcohol, Pure, 200 proof, anhydrous, ≥99.5%
Sigma-Aldrich
Ethanol, ≥99.5%, SAJ super special grade
Sigma-Aldrich
Ethanol, JIS 1000, ≥99.5%, suitable for residue analysis
Sigma-Aldrich
Ethyl alcohol, Pure, 190 proof, meets USP testing specifications
Sigma-Aldrich
Ethyl alcohol, Pure, 190 proof, ACS spectrophotometric grade, 95.0%
Sigma-Aldrich
Allylamine hydrochloride, 98%
Sigma-Aldrich
Glutaraldehyde solution, SAJ first grade, 20.0-26.0%
Sigma-Aldrich
Ethanol, 94.8-95.8%
Sigma-Aldrich
Hydrogen peroxide solution, SAJ first grade, ≥30.0%
Sigma-Aldrich
Sodium chloride solution, 1 M
Sigma-Aldrich
Ethanol, JIS special grade, 94.8-95.8%
Sigma-Aldrich
Sodium chloride, JIS special grade, ≥99.5%
Sigma-Aldrich
Sodium chloride solution, 0.1 M
Sigma-Aldrich
Hydrogen peroxide solution, contains potassium stannate as inhibitor, 30-32 wt. % in water, semiconductor grade, 99.999% trace metals basis
Sigma-Aldrich
Sodium chloride, 99.999% trace metals basis
Sigma-Aldrich
Sodium chloride, random crystals, 99.9% trace metals basis
Sigma-Aldrich
Glutaraldehyde solution, 50 wt. % in H2O
Sigma-Aldrich
Sodium chloride-35Cl, 99 atom % 35Cl
Sigma-Aldrich
Sodium chloride solution, 0.85%
Sigma-Aldrich
Hydrogen peroxide solution, 34.5-36.5%
Sigma-Aldrich
5,5′-Dithiobis(2-nitrobenzoic acid), ReagentPlus®, 99%
Sigma-Aldrich
Sodium chloride, AnhydroBeads, −10 mesh, 99.999% trace metals basis
Sigma-Aldrich
Sodium chloride, BioUltra, Molecular Biology, ≥99.5% (AT)
Sigma-Aldrich
Sodium chloride solution, BioUltra, Molecular Biology, ~5 M in H2O
Sigma-Aldrich
Glutaric dialdehyde solution, 50 wt. % in H2O, FCC