Merck
  • Sox2 antagonizes the Hippo pathway to maintain stemness in cancer cells.
Sox2 antagonizes the Hippo pathway to maintain stemness in cancer cells.
Nature communications (2015-04-04)
Upal Basu-Roy, N Sumru Bayin, Kirk Rattanakorn, Eugenia Han, Dimitris G Placantonakis, Alka Mansukhani, Claudio Basilico
ABSTRACT
The repressive Hippo pathway has a profound tumour suppressive role in cancer by restraining the growth-promoting function of the transcriptional coactivator, YAP. We previously showed that the stem cell transcription factor Sox2 maintains cancer stem cells (CSCs) in osteosarcomas. We now report that in these tumours, Sox2 antagonizes the Hippo pathway by direct repression of two Hippo activators, Nf2 (Merlin) and WWC1 (Kibra), leading to exaggerated YAP function. Repression of Nf2, WWC1 and high YAP expression marks the CSC fraction of the tumor population, while the more differentiated fraction has high Nf2, high WWC1 and reduced YAP expression. YAP depletion sharply reduces CSCs and tumorigenicity of osteosarcomas. Thus, Sox2 interferes with the tumour-suppressive Hippo pathway to maintain CSCs in osteosarcomas. This Sox2-Hippo axis is conserved in other Sox2-dependent cancers such as glioblastomas. Disruption of YAP transcriptional activity could be a therapeutic strategy for Sox2-dependent tumours.
Sigma-Aldrich
MISSION® esiRNA, targeting human NF2
Pricing and availability is not currently available.
Sigma-Aldrich
EGF human, Animal-component free, recombinant, expressed in E. coli, ≥98% (SDS-PAGE), ≥98% (HPLC), suitable for cell culture
Pricing and availability is not currently available.
Sigma-Aldrich
MISSION® esiRNA, targeting mouse Yap1
Pricing and availability is not currently available.
Sigma-Aldrich
EGF from mouse, recombinant, expressed in E. coli, ≥98% (SDS-PAGE), ≥98% (HPLC), suitable for cell culture
Pricing and availability is not currently available.
Sigma-Aldrich
EGF from mouse, Animal-component free, recombinant, expressed in E. coli, ≥98% (SDS-PAGE), ≥98% (HPLC)
Pricing and availability is not currently available.
Sigma-Aldrich
hEGF, EGF, recombinant, expressed in E. coli, lyophilized powder, suitable for cell culture
Pricing and availability is not currently available.
Sigma-Aldrich
EGF from rat, recombinant, expressed in E. coli, ≥98% (SDS-PAGE), ≥98% (HPLC), suitable for cell culture
Pricing and availability is not currently available.
Supelco
Retinol solution, 100 μg/mL ± 25% (Refer to COA) (Ethanol with 0.1% (w/v) BHT), ampule of 1 mL, reference material, Cerilliant®
Pricing and availability is not currently available.
Sigma-Aldrich
MISSION® esiRNA, targeting human SOX2
Pricing and availability is not currently available.
Sigma-Aldrich
MISSION® esiRNA, targeting human YAP1
Pricing and availability is not currently available.
Sigma-Aldrich
MISSION® esiRNA, targeting human YY1AP1
Pricing and availability is not currently available.
Sigma-Aldrich
Epidermal Growth Factor, human, animal component free, EGF, recombinant, expressed in Escherichia coli, >97% (SDS-PAGE)
Pricing and availability is not currently available.