Merck
  • Molecular Basis for the Regulation of Transcriptional Coactivator p300 in Myogenic Differentiation.
Molecular Basis for the Regulation of Transcriptional Coactivator p300 in Myogenic Differentiation.
Scientific reports (2015-09-12)
Jihong Chen, Yingjian Wang, Munerah Hamed, Natascha Lacroix, Qiao Li
ABSTRACT
Skeletal myogenesis is a highly ordered process which specifically depends on the function of transcriptional coactivator p300. Previous studies have established that Akt/protein kinase B (PKB), a positive regulator of p300 in proliferating cells, is also important for proper skeletal muscle development. Nevertheless, it is not clear as to how the p300 is regulated by myogenic signaling events given that both p300 and Akt are involved in many cellular processes. Our studies revealed that the levels of p300 protein are temporally maintained in ligand-enhanced skeletal myocyte development. Interestingly, this maintenance of p300 protein is observed at the stage of myoblast differentiation, which coincides with an increase in Akt phosphorylation. Moreover, regulation of p300 during myoblast differentiation appears to be mediated by Akt signaling. Blunting of p300 impairs myogenic expression and myoblast differentiation. Thus, our data suggests a particular role for Akt in myoblast differentiation through interaction with p300. Our studies also establish the potential of exploiting p300 regulation and Akt activation to decipher the complex signaling cascades involved in skeletal muscle development.
Sigma-Aldrich
MISSION® esiRNA, targeting human NOTCH1
Sigma-Aldrich
Monoclonal Anti-β-Actin antibody produced in mouse, clone AC-15, ascites fluid
Sigma-Aldrich
MISSION® esiRNA, targeting human EP300
Sigma-Aldrich
Dimethyl sulfoxide, JIS special grade, ≥99.0%
Sigma-Aldrich
Dimethyl sulfoxide, SAJ first grade, ≥99.0%
Sigma-Aldrich
Dimethyl sulfoxide, ≥99.5%
Sigma-Aldrich
Dimethyl sulfoxide, ≥99.0%, suitable for absorption spectrum analysis
Sigma-Aldrich
Dimethyl sulfoxide, suitable for HPLC
Sigma-Aldrich
p300 (C1135-2414) human, recombinant, expressed in insect cells, ≥70% (SDS-PAGE)
Sigma-Aldrich
MISSION® esiRNA, targeting mouse Notch1
Sigma-Aldrich
MISSION® esiRNA, targeting mouse Ep300
Sigma-Aldrich
Dimethyl sulfoxide, BioUltra, for molecular biology, ≥99.5% (GC)
Sigma-Aldrich
Dimethyl sulfoxide, anhydrous, ≥99.9%
Sigma-Aldrich
8-Octanoyloxypyrene-1,3,6-trisulfonic acid trisodium salt, suitable for fluorescence, ≥90% (HPCE)
Sigma-Aldrich
Dimethyl sulfoxide, meets EP testing specifications, meets USP testing specifications
Sigma-Aldrich
Dimethyl sulfoxide, for molecular biology
Sigma-Aldrich
Dimethyl sulfoxide, sterile-filtered, BioPerformance Certified, meets EP, USP testing specifications, suitable for hybridoma
Sigma-Aldrich
Dimethyl sulfoxide, ≥99.5% (GC), suitable for plant cell culture
Sigma-Aldrich
Dimethyl sulfoxide, PCR Reagent
Sigma-Aldrich
Dimethyl sulfoxide, Hybri-Max, sterile-filtered, BioReagent, suitable for hybridoma, ≥99.7%
Sigma-Aldrich
Dimethyl sulfoxide, Vetec, reagent grade, 99%