Merck
  • Discovery of syn-/anti-cocaine-N-oxide diastereomers in unwashed postmortem hair via LC-MS-MS.
Discovery of syn-/anti-cocaine-N-oxide diastereomers in unwashed postmortem hair via LC-MS-MS.
Journal of analytical toxicology (2014-05-02)
Christine M Marsh, Lindsey R Crawley, Sarah K Himes, Roman Aranda, Mark L Miller
ABSTRACT
The discovery of two cocaine-N-oxide (CNO) diastereomers, syn- and anti-CNO, is reported for the first time. Prior to this study, only one structural form of CNO was known to exist and has not been analyzed in hair before. CNO is a metabolite of cocaine (COC) and may be considered as an additional biomarker of COC use, along with other known COC metabolites. The analysis of COC in hair for forensic applications is under scrutiny due to the possibility of external contamination. A qualitative liquid chromatography-tandem mass spectrometry method was developed, validated and applied to unwashed postmortem hair samples from drug users. The limit of detection in hair was 8 pg/mg (using 10 mg of unwashed hair) for each CNO diastereomer. The presence of both syn- and anti-forms of CNO was verified in vivo using hair samples collected from known COC-using individuals. Due to the low levels of CNO, it will not always be detectable in COC user hair. In the hair samples analyzed, syn-CNO was detected in more samples than anti-CNO. The stereoselective N-oxidation of COC which favors syn-CNO could have a diagnostic value for COC ingestion determination in hair analysis.
Sigma-Aldrich
Hydrogen peroxide solution, SAJ first grade, ≥30.0%
Sigma-Aldrich
Acetonitrile solution, contains 0.05 % (v/v) trifluoroacetic acid
Sigma-Aldrich
Hydrogen peroxide solution, tested according to Ph. Eur.
Sigma-Aldrich
Hydrogen peroxide solution, contains potassium stannate as inhibitor, 30-32 wt. % in water, semiconductor grade, 99.999% trace metals basis
Supelco
Hydrogen peroxide solution, ≥30%, for trace analysis
Sigma-Aldrich
Hydrogen peroxide solution, 34.5-36.5%
Millipore
Hydrogen peroxide solution, 3%, suitable for microbiology
Sigma-Aldrich
Methanol, ACS reagent, ≥99.8%
Sigma-Aldrich
Methanol, puriss., meets analytical specification of Ph Eur, ≥99.7% (GC)
Sigma-Aldrich
Methanol, ACS spectrophotometric grade, ≥99.9%
Sigma-Aldrich
Methanol, suitable for HPLC, ≥99.9%
Sigma-Aldrich
Methanol, puriss. p.a., ACS reagent, reag. ISO, reag. Ph. Eur., ≥99.8% (GC)
Sigma-Aldrich
Hydrogen peroxide solution, contains inhibitor, 30 wt. % in H2O, meets USP testing specifications
Sigma-Aldrich
Hydrogen peroxide solution, 50 wt. % in H2O, stabilized
Sigma-Aldrich
Methanol, HPLC Plus, ≥99.9%
Sigma-Aldrich
Hydrogen peroxide solution, contains inhibitor, 30 wt. % in H2O, ACS reagent
Sigma-Aldrich
Methanol, Laboratory Reagent, ≥99.6%
Sigma-Aldrich
Methanol, Absolute - Acetone free
Sigma-Aldrich
Hydrogen peroxide solution, contains inhibitor, 35 wt. % in H2O
Sigma-Aldrich
Methanol, suitable for HPLC, gradient grade, ≥99.9%
Sigma-Aldrich
Methanol, BioReagent, ≥99.93%
Supelco
Acetonitrile(Neat), Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
Methanol, ACS reagent, ≥99.8%
Sigma-Aldrich
Methanol, ACS reagent, ≥99.8%
Sigma-Aldrich
Methanol, NMR reference standard
Sigma-Aldrich
Hydrogen peroxide solution, 30 % (w/w) in H2O, contains stabilizer
Sigma-Aldrich
Methanol, JIS special grade, ≥99.8%
Sigma-Aldrich
Methanol, suitable for HPLC
Sigma-Aldrich
Methanol, HPLC Plus, ≥99.9%, poly-coated bottles
Sigma-Aldrich
Acetonitrile solution, contains 0.05 % (w/v) ammonium formate, 5 % (v/v) water, 0.1 % (v/v) formic acid, suitable for HPLC