Merck
  • IL-15 amplifies the pathogenic properties of CD4+CD28- T cells in multiple sclerosis.
IL-15 amplifies the pathogenic properties of CD4+CD28- T cells in multiple sclerosis.
Journal of immunology (Baltimore, Md. : 1950) (2015-01-27)
Bieke Broux, Mark R Mizee, Marjan Vanheusden, Susanne van der Pol, Jack van Horssen, Bart Van Wijmeersch, Veerle Somers, Helga E de Vries, Piet Stinissen, Niels Hellings
ABSTRACT
CD4(+)CD28(-) T cells arise through repeated antigenic stimulation and are present in diseased tissues of patients with various autoimmune disorders, including multiple sclerosis (MS). These cells are believed to have cytotoxic properties that contribute to the pathogenic damaging of the target organ. Endogenous cues that are increased in the diseased tissue may amplify the activity of CD4(+)CD28(-) T cells. In this study, we focused on IL-15, a cytotoxicity-promoting cytokine that is increased in the serum and cerebrospinal fluid of MS patients. Using immunohistochemistry, we demonstrate that IL-15 is mainly produced by astrocytes and infiltrating macrophages in inflammatory lesions of MS patients. Moreover, in vitro transmigration studies reveal that IL-15 selectively attracts CD4(+)CD28(-) T cells of MS patients, but not of healthy individuals. IL-15 further induces the expression of chemokine receptors and adhesion molecules on CD4(+)CD28(-) T cells, as investigated using flow cytometry, resulting in enhanced migration over a monolayer of human brain endothelial cells. Finally, flow cytometric analyses revealed that IL-15 increases the proliferation and production of GM-CSF, expression of cytotoxic molecules (NKG2D, perforin, and granzyme B), and degranulation capacity of CD4(+)CD28(-) T cells. Taken together, these findings indicate that increased peripheral and local levels of IL-15 amplify the pathogenic potential of CD4(+)CD28(-) T cells, thus contributing to tissue damage in MS brain lesions.
Sigma-Aldrich
5(6)-Carboxyfluorescein diacetate N-succinimidyl ester, BioReagent, suitable for fluorescence, ≥90% (HPLC)
Sigma-Aldrich
5-Carboxy-fluorescein diacetate N-succinimidyl ester, for fluorescence, ≥95.0% (HPLC)
Sigma-Aldrich
Phorbol 12-myristate 13-acetate, synthetic, ≥98.0% (TLC)
Supelco
Ethanol solution, certified reference material, 2000 μg/mL in methanol
Sigma-Aldrich
Sodium pyruvate, powder, BioReagent, suitable for cell culture, suitable for insect cell culture, ≥99%
Sigma-Aldrich
Sodium pyruvate, BioXtra, ≥99%
Sigma-Aldrich
Sodium pyruvate, powder, BioXtra, suitable for mouse embryo cell culture
Sigma-Aldrich
Sodium pyruvate, Hybri-Max, powder, suitable for hybridoma
Sigma-Aldrich
Sodium pyruvate, ReagentPlus®, ≥99%
Sigma-Aldrich
Ethanol Fixative 80% v/v, suitable for fixing solution (blood films)
Sigma-Aldrich
Phorbol 12-myristate 13-acetate, ≥99% (TLC), film or powder
Sigma-Aldrich
Ethanol, JIS special grade, 94.8-95.8%
Sigma-Aldrich
Ethanol, JIS first grade, 94.8-95.8%
Supelco
Dehydrated Alcohol, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
Sodium pyruvate, SAJ special grade, ≥95.0%
USP
Dehydrated Alcohol, United States Pharmacopeia (USP) Reference Standard
Sigma-Aldrich
Sodium pyruvate, anhydrous, free-flowing, Redi-Dri, ReagentPlus®, ≥99%
Sigma-Aldrich
Ethyl alcohol, Pure, 190 proof, ACS spectrophotometric grade, 95.0%
Sigma-Aldrich
Ethyl alcohol, Pure, 190 proof, meets USP testing specifications
Supelco
Ethanol standards 10% (v/v), 10 % (v/v) in H2O, analytical standard
Sigma-Aldrich
Ethanol, ≥99.5%, suitable for fluorescence
Sigma-Aldrich
Ethanol, ≥99.5%
Sigma-Aldrich
Ethanol, JIS 300, ≥99.5%, for residue analysis
Sigma-Aldrich
Ethanol, ≥99.5%, suitable for HPLC
Sigma-Aldrich
Ethanol, ≥99.5%, suitable for absorption spectrum analysis
Sigma-Aldrich
Ethanol, ≥99.5%, SAJ super special grade
Sigma-Aldrich
Ethanol, JIS 1000, ≥99.5%, for residue analysis
Sigma-Aldrich
IL-15 human, Animal-component free, recombinant, expressed in E. coli, ≥98% (SDS-PAGE), ≥98% (HPLC), suitable for cell culture
Sigma-Aldrich
Ethanol, purum, fine spirit, denaturated with 4.8% methanol, F25 METHYL1, ~96% (based on denaturant-free substance)
Sigma-Aldrich
Ethanol, ACS reagent, prima fine spirit, without additive, F15 o1