Merck
  • Effects of glucose on cell viability and antioxidant and anti-inflammatory properties of phytochemicals and phytochemically modified membranes.
Effects of glucose on cell viability and antioxidant and anti-inflammatory properties of phytochemicals and phytochemically modified membranes.
The journal of physical chemistry. B (2014-09-24)
Teng Chang, Chandrasekaran Neelakandan, Linda DeFine, Thomas Alexander, Thein Kyu
ABSTRACT
By virtue of antioxidant and anti-inflammable properties, plant-derived phytochemicals such as mangiferin and genistein have attracted considerable attention for functionalization of polymeric hemodialysis (HD) membranes via solution blending. In-vitro dihydrorhodamine (DHR) assay of the genistein-modified membranes revealed drastic reduction in the level of the reactive oxygen species (ROS). In contrast, mangiferin-modified HD membrane manifested the pro-oxidant activity. We suspected that such difference in ROS generation may be attributed to the glucose unit on the xanthone backbone of mangiferin. This hypothesis was confirmed by comparing the ROS levels of genistein versus genistin, and mangiferin versus xanthone and 3,4,5,6-tetrahydroxyxanthone. Phytochemicals without the glucose unit show better antioxidant property related to the glycosides. Anti-inflammatory property was further conducted by measuring the level of TNF-α in blood after contacting with the same selected phytochemicals. Of particular interest is that the glucose unit promotes the generation of TNF-α.
Sigma-Aldrich
Dimethyl sulfoxide, suitable for HPLC
Sigma-Aldrich
Dimethyl sulfoxide, ≥99.5%
Sigma-Aldrich
Dimethyl sulfoxide, JIS special grade, ≥99.0%
Sigma-Aldrich
Dimethyl sulfoxide solution, 50 wt. % in H2O
Sigma-Aldrich
Dimethyl sulfoxide, ≥99.0%, suitable for absorption spectrum analysis
Sigma-Aldrich
Dimethyl sulfoxide, SAJ first grade, ≥99.0%
Sigma-Aldrich
Dimethyl sulfoxide, ≥99.5% (GC), suitable for plant cell culture
Sigma-Aldrich
Dimethyl sulfoxide, suitable for HPLC, ≥99.7%
Sigma-Aldrich
Dimethyl sulfoxide, ReagentPlus®, ≥99.5%
Sigma-Aldrich
Dimethyl sulfoxide, puriss. p.a., ACS reagent, ≥99.9% (GC)
Sigma-Aldrich
Dimethyl sulfoxide, Vetec, reagent grade, 99%
Supelco
Dimethyl sulfoxide, for inorganic trace analysis, ≥99.99995% (metals basis)
Sigma-Aldrich
Dimethyl sulfoxide, ACS reagent, ≥99.9%
Sigma-Aldrich
Dimethyl sulfoxide, for molecular biology
Sigma-Aldrich
Dimethyl sulfoxide, anhydrous, ≥99.9%
Sigma-Aldrich
Dimethyl sulfoxide, sterile-filtered, BioPerformance Certified, meets EP, USP testing specifications, suitable for hybridoma
Supelco
Dimethyl sulfoxide, analytical standard
Sigma-Aldrich
Dimethyl sulfoxide, Hybri-Max, sterile-filtered, BioReagent, suitable for hybridoma, ≥99.7%
Sigma-Aldrich
Dimethyl sulfoxide, BioUltra, for molecular biology, ≥99.5% (GC)
Sigma-Aldrich
Dimethyl sulfoxide, PCR Reagent
Sigma-Aldrich
Dimethyl sulfoxide, meets EP testing specifications, meets USP testing specifications
USP
Dimethyl sulfoxide, United States Pharmacopeia (USP) Reference Standard
Dimethyl sulfoxide, European Pharmacopoeia (EP) Reference Standard
Sigma-Aldrich
Dimethyl sulfoxide, puriss. p.a., dried, ≤0.02% water
Sigma-Aldrich
Xanthone, 97%
Sigma-Aldrich
7-Aminoactinomycin D, ~97% (HPLC), powder
Sigma-Aldrich
Dihydrorhodamine 123, ≥95%
USP
Xanthone, United States Pharmacopeia (USP) Reference Standard