Merck
  • Biopharmaceutical and pharmacokinetic characterization of asiatic acid in Centella asiatica as determined by a sensitive and robust HPLC-MS method.
Biopharmaceutical and pharmacokinetic characterization of asiatic acid in Centella asiatica as determined by a sensitive and robust HPLC-MS method.
Journal of ethnopharmacology (2015-01-18)
Yongfang Yuan, Hai Zhang, Fengfeng Sun, Sen Sun, Zhenyu Zhu, Yifeng Chai
ABSTRACT
Asiatic acid is one of the main components in the herb Centella asiatica, which is a well-known herbal medicine for its excellent pharmacological effects. To enhance the development potentials of asiatic acid as a chemopreventative agent, there is a great need to further understand its biopharmaceutical and pharmacokinetic properties. The aim of this research is to clarify the mechanisms of absorption and metabolism of asiatic acid, and explore its biopharmaceutical and pharmacokinetic properties in rats by using a sensitive and robust HPLC-MS method. Male Sprague-Dawley rats were randomly assigned to 2 groups and administered with asiatic acid by oral and intravenous administration. Plasma concentrations of asiatic acid were determined at designated points and main pharmacokinetic parameters were estimated. The absorption of asiatic acid was investigated by using Caco-2 cell line absorption model in vitro and rat intestinal perfusion model in situ. The metabolic rate of asiatic acid was investigated by incubating it in rat liver microsome system in vitro. In addition, the solubility of asiatic acid in aqueous solution was also determined by using HPLC-MS method. The absolute oral bioavailability of asiatic acid is 16.25%. It was found that the permeability of asiatic acid is more than 10(-5) in the Caco-2 cell monolayer and rat intestinal perfusion model, and its main absorption region is the jejunum in rats. The metabolic rate of asiatic acid in rat liver microsomes, t1/2, is 9.493min, which shows that asiatic acid can be metabolized rapidly. The solubility of aisiatic acid was 0.1583mgmL(-1), and its poor solubility will result in low bioavailability. The asiatic acid in a variety of matrixes was analyzed by using a sensitive and specific HPLC-MS method, and its absolute oral bioavailability in rats was very low. Asiatic acid can be metabolized rapidly in rat liver microsomes, and has good permeability across Caco-2 monolayer cell and rat intestine perfusion. It can be deduced that the low bioavailability of asiatic acid results from poor solubility and rapid metabolism.
Sigma-Aldrich
Acetonitrile solution, contains 0.05 % (v/v) trifluoroacetic acid
Sigma-Aldrich
Acetonitrile solution, contains 0.1 % (v/v) trifluoroacetic acid, suitable for HPLC
Sigma-Aldrich
Acetonitrile solution, contains 0.1 % (v/v) formic acid, suitable for HPLC
Sigma-Aldrich
Acetonitrile solution, contains 0.05 % (w/v) ammonium formate, 5 % (v/v) water, 0.1 % (v/v) formic acid, suitable for HPLC
Sigma-Aldrich
Acetonitrile solution, contains 10.0% acetone, 0.05% formic acid, 40.0% 2-propanol
Supelco
Residual Solvent - Acetonitrile, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
Acetonitrile, HPLC Plus, ≥99.9%, poly-coated bottles
Sigma-Aldrich
Acetonitrile, for residue analysis, JIS 5000
Sigma-Aldrich
Acetonitrile, for chromatography
Sigma-Aldrich
Acetonitrile, SAJ first grade, ≥99.0%
Sigma-Aldrich
Acetonitrile, JIS special grade, ≥99.5%
Sigma-Aldrich
Formic acid, JIS special grade, ≥98.0%
Sigma-Aldrich
Acetonitrile, ≥99.8%, for residue analysis, JIS 300
Sigma-Aldrich
Acetonitrile, electronic grade, 99.999% trace metals basis
Sigma-Aldrich
Asiatic acid, ≥98% (HPLC), from Centella asiatica
Supelco
Acetonitrile, analytical standard
Supelco
Acetonitrile, HPLC grade, ≥99.93%
Sigma-Aldrich
Acetonitrile, ≥99.5%, ACS reagent
Sigma-Aldrich
Acetonitrile, anhydrous, 99.8%
Sigma-Aldrich
Asiatic acid, 97%
Sigma-Aldrich
Acetonitrile, suitable for HPLC, gradient grade, ≥99.9%
Sigma-Aldrich
Formic acid, reagent grade, ≥95%
Sigma-Aldrich
Acetonitrile, biotech. grade, ≥99.93%
Sigma-Aldrich
Formic acid, ≥95%, FCC, FG
Sigma-Aldrich
Acetonitrile, ≥99.8%, suitable for HPLC
Sigma-Aldrich
Acetonitrile, HPLC Plus, ≥99.9%
Supelco
Asiatic acid, analytical standard
Sigma-Aldrich
Acetonitrile, ≥99.8%, for residue analysis, JIS 1000
Sigma-Aldrich
Acetonitrile, ACS reagent, ≥99.5%
Sigma-Aldrich
Formic acid, puriss. p.a., ACS reagent, reag. Ph. Eur., ≥98%