Merck
  • Patterns of genome evolution that have accompanied host adaptation in Salmonella.
Patterns of genome evolution that have accompanied host adaptation in Salmonella.
Proceedings of the National Academy of Sciences of the United States of America (2014-12-24)
Gemma C Langridge, Maria Fookes, Thomas R Connor, Theresa Feltwell, Nicholas Feasey, Bryony N Parsons, Helena M B Seth-Smith, Lars Barquist, Anna Stedman, Tom Humphrey, Paul Wigley, Sarah E Peters, Duncan J Maskell, Jukka Corander, Jose A Chabalgoity, Paul Barrow, Julian Parkhill, Gordon Dougan, Nicholas R Thomson
ABSTRACT
Many bacterial pathogens are specialized, infecting one or few hosts, and this is often associated with more acute disease presentation. Specific genomes show markers of this specialization, which often reflect a balance between gene acquisition and functional gene loss. Within Salmonella enterica subspecies enterica, a single lineage exists that includes human and animal pathogens adapted to cause infection in different hosts, including S. enterica serovar Enteritidis (multiple hosts), S. Gallinarum (birds), and S. Dublin (cattle). This provides an excellent evolutionary context in which differences between these pathogen genomes can be related to host range. Genome sequences were obtained from ∼ 60 isolates selected to represent the known diversity of this lineage. Examination and comparison of the clades within the phylogeny of this lineage revealed signs of host restriction as well as evolutionary events that mark a path to host generalism. We have identified the nature and order of events for both evolutionary trajectories. The impact of functional gene loss was predicted based upon position within metabolic pathways and confirmed with phenotyping assays. The structure of S. Enteritidis is more complex than previously known, as a second clade of S. Enteritidis was revealed that is distinct from those commonly seen to cause disease in humans or animals, and that is more closely related to S. Gallinarum. Isolates from this second clade were tested in a chick model of infection and exhibited a reduced colonization phenotype, which we postulate represents an intermediate stage in pathogen-host adaptation.
Sigma-Aldrich
Ethanolamine, JIS special grade, ≥99.0%
Sigma-Aldrich
Ethanolamine, SAJ first grade, ≥99.0%
Supelco
Propylene glycol, Pharmaceutical Secondary Standard; Certified Reference Material
Aucubin, primary reference standard
Sigma-Aldrich
Ethanolamine, liquid, BioReagent, suitable for cell culture, ≥98%
Sigma-Aldrich
Ethanolamine, ≥99%
Sigma-Aldrich
Ethanolamine, ≥98%
Sigma-Aldrich
Ethanolamine, ACS reagent, ≥99.0%
Sigma-Aldrich
Ethanolamine, puriss. p.a., ACS reagent, ≥99.0% (GC/NT)
Supelco
Ethanolamine, analytical standard
Sigma-Aldrich
Ethanolamine, purified by redistillation, ≥99.5%
Sigma-Aldrich
Propylene Glycol, meets USP testing specifications
Sigma-Aldrich
Propylene glycol, ≥99.5% (GC), FCC, FG
USP
Propylene glycol, United States Pharmacopeia (USP) Reference Standard
Supelco
Aucubin, analytical standard
Propylene glycol, European Pharmacopoeia (EP) Reference Standard
Sigma-Aldrich
1,2-Propanediol, meets analytical specification of Ph. Eur., BP, USP, ≥99.5%
Sigma-Aldrich
1,2-Propanediol, puriss. p.a., ACS reagent, ≥99.5% (GC)
Supelco
1,2-Propanediol, analytical standard
Sigma-Aldrich
1,2-Propanediol, ACS reagent, ≥99.5%
Sigma-Aldrich
1,2-Propanediol, ReagentPlus®, 99%
Sigma-Aldrich
1,2-Propanediol, SAJ first grade, ≥99.0%
Sigma-Aldrich
1,2-Propanediol, SAJ special grade, ≥99.0%
Sigma-Aldrich
1,2-Propanediol, tested according to Ph. Eur.
Trolamine impurity A, European Pharmacopoeia (EP) Reference Standard