Merck
  • Composition and antiproliferative effect of essential oil of Origanum vulgare against tumor cell lines.
Composition and antiproliferative effect of essential oil of Origanum vulgare against tumor cell lines.
Journal of medicinal food (2014-09-18)
Karine Rech Begnini, Fernanda Nedel, Rafael Guerra Lund, Pedro Henrique de Azambuja Carvalho, Maria Regina Alves Rodrigues, Fátima Tereza Alves Beira, Francisco Augusto Burkert Del-Pino
ABSTRACT
Cancer is a leading cause of death and is responsible for one in eight deaths worldwide. The use of herbs as complementary medicine for cancer, especially advanced cancer, has recently increased. The aim of this study was to evaluate in vitro, the antiproliferative effect of Origanum vulgare against human breast adenocarcinoma (MCF-7), and human colon adenocarcinoma (HT-29). The essential oil (EO) was extracted from a bought amount of O. vulgare dried leaves and analyzed in a gas chromatograph interfaced with a mass selective detector. The cytotoxicity test was performed by sulforhodamine B assay. The results show that the EO is composed mostly of 4-terpineol and induces a high cytotoxicity effect in HT-29. In the MCF-7 cell line the EO was less effective. In conclusion, this study showed that O. vulgare main component is 4-terpineol and was effective in inducing cancer cell growth inhibition.
Product Description
Supelco
1,8-Cineole, primary reference standard
Sigma-Aldrich
Dichloromethane, JIS special grade, ≥99.0%
Sigma-Aldrich
Dichloromethane, SAJ first grade, ≥99.0%
Supelco
Thymol, Pharmaceutical Secondary Standard; Certified Reference Material
Supelco
Camphene, analytical standard
Supelco
Dichloromethane solution, contains 10 % (v/v) methanol
Sigma-Aldrich
Thymol, ≥98.5%
Sigma-Aldrich
Sigma 7-9®, ≥99% (titration), crystalline
Sigma-Aldrich
Terpineol, mixture of isomers
Supelco
Dichloromethane, Selectophore, for ion-selective electrodes, ≥99.5%
Sigma-Aldrich
Thymol, meets analytical specification of Ph. Eur., BP, NF, 99-101%
Supelco
Dichloromethane, ≥99.9%
Supelco
(−)-Linalool, analytical standard
Supelco
Dichloromethane, analytical standard
Sigma-Aldrich
Dichloromethane, anhydrous, ≥99.8%, contains 40-150 ppm amylene as stabilizer
Thymol, European Pharmacopoeia (EP) Reference Standard
Sigma-Aldrich
Terpineol, SAJ first grade, ≥97.0%
Camphene, primary reference standard
Sigma-Aldrich
Dichloromethane, ACS reagent, ≥99.5%, contains 40-150 ppm amylene as stabilizer
SAFC
Tromethamine
Sigma-Aldrich
Dichloromethane, contains 40-150 ppm amylene as stabilizer, ACS reagent, ≥99.5%
Sigma-Aldrich
Dichloromethane, puriss., meets analytical specification of Ph. Eur., NF, ≥99% (GC)
Sigma-Aldrich
Dichloromethane, suitable for HPLC, ≥99.8%, contains amylene as stabilizer
Sigma-Aldrich
Thymol, FCC, FG
Sigma-Aldrich
Camphene, 95%
Supelco
Thymol, TraceCERT®, Manufactured by: Sigma-Aldrich Production GmbH, Switzerland
Supelco
Methylene Chloride, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
Dichloromethane, suitable for HPLC, ≥99.9%, contains 40-150 ppm amylene as stabilizer
Sigma-Aldrich
Dichloromethane, ACS reagent, ≥99.5%, contains 40-150 ppm amylene as stabilizer
Sigma-Aldrich
Dichloromethane, biotech. grade, 99.9%, contains 40-150 ppm amylene as stabilizer