Skip to Content
Merck
All Photos(2)

Key Documents

RES3193T-A7

SAFC

Tromethamine

Synonym(s):

Tromethamine, Tris

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C4H11NO3
CAS Number:
Molecular Weight:
121.14
EC Number:
MDL number:
UNSPSC Code:
12352116
NACRES:
NA.21

biological source

synthetic

Quality Level

form

powder

technique(s)

cell culture | mammalian: suitable

impurities

Endotoxin, microbial, and heavy metals; tested

suitability

suitable for manufacturing use

foreign activity

Cytotoxicity, DNase, NICKase, RNase, and Protease; tested

SMILES string

NC(CO)(CO)CO

InChI

1S/C4H11NO3/c5-4(1-6,2-7)3-8/h6-8H,1-3,5H2

InChI key

LENZDBCJOHFCAS-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Our SAFC® portfolio of high-quality raw materials for use in biopharmaceutical processing withstands strict quality control procedures plus the documentation and expertise to help our customers meet requirements as defined by the M-Clarity Program.

M-Clarity Program

Buffer quality is vital for the success of biopharmaceutical processes, because buffers are indispensable in nearly every production step.

Our broad portfolio of buffer materials manufactured under appropriate controls is tailored to your needs. Ranging from non-GMP grades for low-risk application, to IPEC-PQG GMP for higher-risk applications, we have products covering all your manufacturing needs.

Application

Tromethamine is a commonly used biological buffer which is also referred to as Tris or Trizma. Tris as a buffer has very broad usage in diagnostics and downstream purifications. The pKa of Tris is 7.77 which corresponds closely to a physiological pH of 7.36.

Tromethamine is used in biopharmaceutical buffer formulations (both upstream and downstream), electrophoresis separations and diagnostic reagent formulations. Tromethamine is especially useful in downstream purification processes.

Packaging

Product is available in the following package sizes:
RES3193T-A701X: 100 gm container
RES3193T-A703X: 5 kg container
RES3193T-A705X: 25 kg container
RES3193T-A706X: 50 kg container

Legal Information

SAFC is a registered trademark of Merck KGaA, Darmstadt, Germany

replaced by

Product No.
Description
Pricing

Storage Class Code

11 - Combustible Solids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Sylvain Clède et al.
Chemical communications (Cambridge, England), 48(62), 7729-7731 (2012-06-29)
A rhenium tris-carbonyl derivative has been designed to couple infrared and luminescent detection in cells. Both spectroscopies are consistent with one another; they point out the reliability of the present SCoMPI (for Single Core Multimodal Probe for Imaging) for bimodal
Corey A Baron et al.
Magnetic resonance in medicine, 73(3), 1075-1084 (2014-04-12)
An acquisition method that does not increase scan time or specific absorption rate is investigated for reducing the deleterious effects of cerebrospinal fluid (CSF) partial volume effects on diffusion tensor imaging (DTI) tractography. It is based on using a shorter
Filippo Bertoli et al.
Small (Weinheim an der Bergstrasse, Germany), 10(16), 3307-3315 (2014-04-17)
Nanoparticles in contact with cells and living organisms generate quite novel interactions at the interface between the nanoparticle surface and the surrounding biological environment. However, a detailed time resolved molecular level description of the evolving interactions as nanoparticles are internalized
D R Brocks et al.
Clinical pharmacokinetics, 23(6), 415-427 (1992-12-01)
Ketorolac is a new chiral nonsteroidal anti-inflammatory drug (NSAID) which is marketed for analgesia as the racemate. The drug is administered as the water soluble tromethamine salt and is available in tablets or as an intramuscular injection. The absorption of
Noelia Purroy et al.
Oncotarget, 6(10), 7632-7643 (2014-12-30)
Chronic lymphocytic leukemia (CLL) cells residing in the bone marrow (BM) and in secondary lymphoid tissues receive survival and proliferative signals from the microenvironment, resulting in persistence of residual disease after treatment. In this study, we characterized primary CLL cells

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service