Merck
  • Transforming growth factor-β-activated kinase 1 resistance limits glucocorticoid responsiveness to Toll-like receptor 4-mediated inflammation.
Transforming growth factor-β-activated kinase 1 resistance limits glucocorticoid responsiveness to Toll-like receptor 4-mediated inflammation.
Immunology (2014-12-19)
Fansheng Kong, Gloria Laryea, Zhiwei Liu, Sandip Bhattacharyya
ABSTRACT
Glucocorticoids (GC) are among the most effective anti-inflammatory drugs, but are often associated with serious adverse effects or inadequate therapeutic responses. Here, we use activation of different Toll-like receptors (TLRs) by their respective ligands to evaluate context-specific GC sensitivity in the macrophage. Recruitment and activation of transforming growth factor-β-activated kinase 1 (TAK1), downstream of TLR engagement, is crucial in activating multiple inflammatory pathways, and contributes to inflammatory disorders. We hypothesize that GC exert anti-inflammatory effects through regulation of TAK1. Both in vivo and in vitro, in comparison to other TLRs, there was limited GC potency in restricting TLR4 ligand-mediated secretion of interleukin-6, tumour necrosis factor-α and interleukin-12. Also, we found that inactivation of TAK1 both in vivo and in vitro strongly inhibits TLR4-induced inflammation-associated genes beyond the suppressive effects from GC treatment. However, there was no effect of TAK1 inactivation on GC inhibition of TLR3- or TLR9-initiated inflammatory actions. Together, our findings demonstrate that GC resistance for TAK1 activation associated with TLR4 engagement may be an important contributor to GC resistance in inflammatory disorders.
Sigma-Aldrich
Sodium chloride solution, 0.1 M
Sigma-Aldrich
Sodium chloride, JIS special grade, ≥99.5%
Sigma-Aldrich
Sodium chloride solution, 1 M
Sigma-Aldrich
Sodium chloride, BioPerformance Certified, ≥99% (titration), suitable for insect cell culture, suitable for plant cell culture
Sigma-Aldrich
Sodium chloride solution, 0.85%
Sigma-Aldrich
Sodium chloride-35Cl, 99 atom % 35Cl
Sigma-Aldrich
Sodium chloride, random crystals, optical grade, 99.9% trace metals basis
Sigma-Aldrich
Sodium chloride, BioUltra, for molecular biology, ≥99.5% (AT)
Sigma-Aldrich
Sodium chloride, tested according to Ph. Eur.
Supelco
Corticosterone, VETRANAL®, analytical standard
Supelco
Sodium chloride, reference material for titrimetry, certified by BAM, >99.5%
Sigma-Aldrich
Corticosterone, ≥98.5% (HPLC)
Sigma-Aldrich
Sodium chloride, 99.999% trace metals basis
Sigma-Aldrich
Sodium chloride solution, 0.9% in water, BioXtra, suitable for cell culture
Sigma-Aldrich
Sodium chloride, BioReagent, suitable for cell culture, suitable for insect cell culture, suitable for plant cell culture, ≥99%
Sigma-Aldrich
Sodium chloride, for molecular biology, DNase, RNase, and protease, none detected, ≥99% (titration)
Sigma-Aldrich
Sodium chloride, BioXtra, ≥99.5% (AT)
Sigma-Aldrich
Sodium chloride solution, 5 M
Sigma-Aldrich
Sodium chloride, tablet
Sigma-Aldrich
Sodium chloride, meets analytical specification of Ph. Eur., BP, USP, 99.0-100.5%
Sigma-Aldrich
Sodium chloride, SAJ first grade, ≥99.0%
Sigma-Aldrich
Corticosterone, ≥92%
Sigma-Aldrich
Sodium chloride solution, 5 M in H2O, BioReagent, for molecular biology, suitable for cell culture
Sigma-Aldrich
Sodium chloride solution, BioUltra, for molecular biology, ~5 M in H2O
Sigma-Aldrich
Sodium chloride, AnhydroBeads, −10 mesh, 99.999% trace metals basis
SAFC
Sodium chloride solution, 5 M
Dexamethasone for peak identification, European Pharmacopoeia (EP) Reference Standard
Supelco
Sodium chloride, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
Ethylenediaminetetraacetic acid, BioUltra, ≥99.0% (KT)
Sigma-Aldrich
Dexamethasone, tested according to Ph. Eur.