Merck
  • Marginally designed new profen analogues have the potential to inhibit cyclooxygenase enzymes.
Marginally designed new profen analogues have the potential to inhibit cyclooxygenase enzymes.
Archiv der Pharmazie (2015-01-13)
Hayrettin O Gülcan, Serdar Ünlü, Anatoli Dimoglo, Yasemin Şahin, İlker Esiringu, Tuba Erçetin, Demet Öz, Mustafa F Şahin
ABSTRACT
The current structure-activity relationship of profens (i.e., 2-arylpropionic acid derivatives, a class of non-steroidal anti-inflammatory drugs) discusses the importance of α-monomethyl substitution on these compounds, since the activities obtained through their corresponding arylacetic acid derivatives (i.e., α-demethylated derivatives) or α,α-dimethyl-substituted compounds are less than what is observed for the parent profens. Unfortunately, this implies a generalization in structure-activity relationships of profens in such a way that a mono-(non-methyl)alkyl group or dialkyl substituent replaced at the α-position of a profen analogue results in abolished activity. Therefore, within this study, we aimed to question this generalization employing ibuprofen, flurbiprofen, and naproxen as model compounds. A series of α-(non-methyl)alkyl-substituted ibuprofen and flurbiprofen analogues as well as α,α-dialkyl-substituted ibuprofen, flurbiprofen, and naproxen derivatives were synthesized and screened for their potential to inhibit cyclooxygenase enzymes. In addition, since profens have negligible potential to inhibit lipoxygenase enzymes, the effect of such derivatization was also questioned in lipoxygenase inhibition assays. The findings only partially agreed with the current structure-activity approach of profens and the activity results of some compounds were found as beyond ordinary.
Sigma-Aldrich
Acetonitrile, suitable for residue analysis, JIS 5000
Sigma-Aldrich
Acetonitrile, suitable for chromatography
Supelco
5α-Androstan-17β-ol-3-one, VETRANAL®, analytical standard
Sigma-Aldrich
Acetonitrile, ≥99.8%, suitable for residue analysis, JIS 300
Sigma-Aldrich
Acetonitrile, ≥99.8%, suitable for residue analysis, JIS 1000
Sigma-Aldrich
Acetonitrile, JIS special grade, ≥99.5%
Sigma-Aldrich
Acetonitrile, ≥99.8%, suitable for HPLC
Sigma-Aldrich
Acetonitrile, SAJ first grade, ≥99.0%
Sigma-Aldrich
Acetonitrile, HPLC Plus, ≥99.9%, poly-coated bottles
Millipore
Bifido Selective Supplement B, suitable for microbiology
Supelco
Acetonitrile, HPLC grade, ≥99.93%
Sigma-Aldrich
5α-Androstan-17β-ol-3-one, ≥97.5%
Sigma-Aldrich
3-Methoxybenzyl bromide, 98%
Sigma-Aldrich
Acetonitrile, ≥99.5%, ACS reagent
Sigma-Aldrich
Acetonitrile, anhydrous, 99.8%
Supelco
Acetonitrile, analytical standard
Sigma-Aldrich
1-Bromo-3-phenylpropane, 98%
Sigma-Aldrich
Acetonitrile, electronic grade, 99.999% trace metals basis
Sigma-Aldrich
Acetonitrile, ReagentPlus®, 99%
Sigma-Aldrich
Acetonitrile, suitable for HPLC, gradient grade, ≥99.9%
Sigma-Aldrich
Acetonitrile, suitable for HPLC-GC, ≥99.8% (GC)
Sigma-Aldrich
Acetonitrile, suitable for HPLC, gradient grade, ≥99.9%
Sigma-Aldrich
Acetonitrile, HPLC Plus, ≥99.9%
Sigma-Aldrich
Acetonitrile, ACS reagent, ≥99.5%
Sigma-Aldrich
Acetonitrile, biotech. grade, ≥99.93%
Sigma-Aldrich
Acetonitrile, suitable for DNA synthesis, ≥99.9% (GC)
Sigma-Aldrich
Ultrapure Acetonitrile
USP
Glacial acetic acid, United States Pharmacopeia (USP) Reference Standard
USP
Residual Solvent Class 2 - Acetonitrile, United States Pharmacopeia (USP) Reference Standard
Sigma-Aldrich
Acetonitrile solution, contains 10.0% acetone, 0.05% formic acid, 40.0% 2-propanol