Merck
  • Autotaxin produced by stromal cells promotes LFA-1-independent and Rho-dependent interstitial T cell motility in the lymph node paracortex.
Autotaxin produced by stromal cells promotes LFA-1-independent and Rho-dependent interstitial T cell motility in the lymph node paracortex.
Journal of immunology (Baltimore, Md. : 1950) (2014-06-18)
Tomoya Katakai, Naoyuki Kondo, Yoshihiro Ueda, Tatsuo Kinashi
ABSTRACT
T cells exhibit high-speed migration within the paracortical T zone of lymph nodes (LNs) as they scan cognate Ags displayed by dendritic cells in the tissue microenvironment supported by the network of stromal cells. Although intranodal T cell migration is controlled in part by chemokines and LFA-1/ICAM-1, the mechanisms underlying their migratory activity independent of these factors remain to be elucidated. In this study, we show that LN stromal cells constitutively express autotaxin (ATX), an ectoenzyme that is important for the generation of lysophosphatidic acid (LPA). Importantly, CCL21(+) stromal cells in the T zone produced and immobilized ATX on their cell surface. Two-photon imaging using LN tissue slices revealed that pharmacological inhibition of ATX or LPA receptors significantly reduced T cell migration, and this was further exacerbated by blockage of Gαi signaling or LFA-1. Therefore, T cell motility mediated by the ATX-LPA axis was independent of Gαi and LFA-1. LPA induced slow intermittent movement of T cells in vitro in a LFA-1-independent manner and enhanced CCL21-induced migration. Moreover, LPA and CCL21 cooperatively augmented RhoA activity in T cells, which was necessary for efficient intranodal T cell migration via the downstream ROCK-myosin II pathway. Taken together, T zone stromal cells control optimal migratory behavior of T cells via multiple signaling cues mediated by chemokines and ATX/LPA.
Sigma-Aldrich
Acetone, suitable for HPLC
Sigma-Aldrich
Acetone, for residue analysis, JIS 5000
Sigma-Aldrich
Acetone, ≥99.5%, for residue analysis
Sigma-Aldrich
Acetone, for residue analysis, ≥99.5%
Sigma-Aldrich
Acetone, SAJ first grade, ≥99.0%
Sigma-Aldrich
Acetone, JIS special grade, ≥99.5%
Supelco
Acetone, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
Acetone, for chromatography, ≥99.8%
Sigma-Aldrich
Acetone, suitable for HPLC, ≥99.9%
Sigma-Aldrich
Acetone, natural, ≥97%
Supelco
Acetone, analytical standard
Sigma-Aldrich
HEPES, BioUltra, for molecular biology, ≥99.5% (T)
Sigma-Aldrich
Acetone, ≥99%, meets FCC analytical specifications
Sigma-Aldrich
HEPES, BioXtra, suitable for mouse embryo cell culture, ≥99.5% (titration)
Sigma-Aldrich
HEPES, BioXtra, pH 5.0-6.5 (1 M in H2O), ≥99.5% (titration)
Sigma-Aldrich
HEPES, BioPerformance Certified, ≥99.5% (titration), suitable for cell culture
USP
Acetone, United States Pharmacopeia (USP) Reference Standard
Sigma-Aldrich
HEPES, ≥99.5% (titration)
Supelco
HEPES, Pharmaceutical Secondary Standard; Certified Reference Material
SAFC
HEPES
SAFC
HEPES
Sigma-Aldrich
ANTI-CD106 (CENTER) antibody produced in rabbit, purified immunoglobulin, buffered aqueous solution
Sigma-Aldrich
HEPES, anhydrous, free-flowing, Redi-Dri, ≥99.5%
Sigma-Aldrich
Acetone, Laboratory Reagent, ≥99.5%
Sigma-Aldrich
Acetone, ACS reagent, ≥99.5%
Sigma-Aldrich
Acetone, histological grade, ≥99.5%
Sigma-Aldrich
Acetone, puriss. p.a., ACS reagent, reag. ISO, reag. Ph. Eur., ≥99.5% (GC)
Sigma-Aldrich
Acetone, ACS reagent, ≥99.5%
Sigma-Aldrich
Acetone, suitable for HPLC, ≥99.8%
Sigma-Aldrich
Acetone, suitable for HPLC, ≥99.9%