Merck
  • iRegulon: from a gene list to a gene regulatory network using large motif and track collections.
iRegulon: from a gene list to a gene regulatory network using large motif and track collections.
PLoS computational biology (2014-07-25)
Rekin's Janky, Annelien Verfaillie, Hana Imrichová, Bram Van de Sande, Laura Standaert, Valerie Christiaens, Gert Hulselmans, Koen Herten, Marina Naval Sanchez, Delphine Potier, Dmitry Svetlichnyy, Zeynep Kalender Atak, Mark Fiers, Jean-Christophe Marine, Stein Aerts
ABSTRACT
Identifying master regulators of biological processes and mapping their downstream gene networks are key challenges in systems biology. We developed a computational method, called iRegulon, to reverse-engineer the transcriptional regulatory network underlying a co-expressed gene set using cis-regulatory sequence analysis. iRegulon implements a genome-wide ranking-and-recovery approach to detect enriched transcription factor motifs and their optimal sets of direct targets. We increase the accuracy of network inference by using very large motif collections of up to ten thousand position weight matrices collected from various species, and linking these to candidate human TFs via a motif2TF procedure. We validate iRegulon on gene sets derived from ENCODE ChIP-seq data with increasing levels of noise, and we compare iRegulon with existing motif discovery methods. Next, we use iRegulon on more challenging types of gene lists, including microRNA target sets, protein-protein interaction networks, and genetic perturbation data. In particular, we over-activate p53 in breast cancer cells, followed by RNA-seq and ChIP-seq, and could identify an extensive up-regulated network controlled directly by p53. Similarly we map a repressive network with no indication of direct p53 regulation but rather an indirect effect via E2F and NFY. Finally, we generalize our computational framework to include regulatory tracks such as ChIP-seq data and show how motif and track discovery can be combined to map functional regulatory interactions among co-expressed genes. iRegulon is available as a Cytoscape plugin from http://iregulon.aertslab.org.
Sigma-Aldrich
Sodium pyruvate, SAJ special grade, ≥95.0%
Pricing and availability is not currently available.
Sigma-Aldrich
Sodium pyruvate, powder, BioReagent, suitable for cell culture, suitable for insect cell culture, ≥99%
Pricing and availability is not currently available.
Sigma-Aldrich
Sodium pyruvate, Hybri-Max, powder, suitable for hybridoma
Pricing and availability is not currently available.
Sigma-Aldrich
Sodium pyruvate, BioXtra, ≥99%
Pricing and availability is not currently available.
Sigma-Aldrich
Sodium pyruvate, powder, BioXtra, suitable for mouse embryo cell culture
Pricing and availability is not currently available.
Sigma-Aldrich
Sodium pyruvate, ReagentPlus®, ≥99%
Pricing and availability is not currently available.
Sigma-Aldrich
Sodium pyruvate, anhydrous, free-flowing, Redi-Dri, ReagentPlus®, ≥99%
Pricing and availability is not currently available.