Merck
  • Fed and fasted state gastro-intestinal in vitro lipolysis: In vitro in vivo relations of a conventional tablet, a SNEDDS and a solidified SNEDDS.
Fed and fasted state gastro-intestinal in vitro lipolysis: In vitro in vivo relations of a conventional tablet, a SNEDDS and a solidified SNEDDS.
European journal of pharmaceutical sciences : official journal of the European Federation for Pharmaceutical Sciences (2013-09-24)
Philip Carsten Christophersen, Martin Lau Christiansen, Rene Holm, Jakob Kristensen, Jette Jacobsen, Bertil Abrahamsson, Anette Müllertz
ABSTRACT
The present study aims at evaluating the ability of a gastro-intestinal in vitro lipolysis model to predict the performance of two lipid formulations and a conventional tablet containing a poorly soluble drug, cinnarizine, in dogs, both in the fasted and fed state. A self-nano-emulsifying drug delivery system (SNEDDS) was either dosed in a hard gelatin capsule (SNEDDS-C) or loaded onto a porous tablet core (SNEDDS-T) and compared to a marketed conventional tablet (Conv) in an in vitro lipolysis model. The model simulates the digestion in the stomach and intestine during either the fasted or the fed state. Whole fat milk (3.5%) was used in the fed state model to mimic the dynamic lipolysis events after ingestion of food. The results were compared to a dog study published in this issue. In the fasted state in vitro lipolysis model, the amount of solubilized cinnarizine decreased in the order SNEDDS-C>SNEDDS-T>Conv, which correlated well with the in vivo bioavailability. In the fed state in vitro lipolysis model, cinnarizine was solubilized to the same degree for all formulations. Compared to the fasted state model, only the performance of the conventional tablet was improved, indicating food effect. This correlated with the in vivo study, where the tablet was the only formulation with a significant food effect. The fasted state model correlated well with the in vivo results and although the fed state model did not accurately predict the fed state in vivo results, it could predict which formulation that would exhibit a food effect.
Sigma-Aldrich
Oleic acid, meets analytical specification of Ph, Eur., 65.0-88.0% (GC)
Sigma-Aldrich
Oleic acid, technical grade, 90%
Sigma-Aldrich
Oleic acid, natural, FCC
Supelco
Oleic acid, analytical standard
Sigma-Aldrich
Oleic acid, ≥99% (GC)
Sigma-Aldrich
Oleic acid, BioReagent, suitable for cell culture
Sigma-Aldrich
4-Bromophenylboronic acid, ≥95.0%
Sigma-Aldrich
Oleic acid, SAJ first grade, ≥70.0%
Supelco
Oleic acid, Selectophore, ≥99%
Oleic acid, European Pharmacopoeia (EP) Reference Standard
Sigma-Aldrich
Magnesium stearate, technical grade
Sigma-Aldrich
Magnesium stearate, puriss., meets analytical specification of Ph. Eur., BP, ≥90% stearic and palmitic acid basis, ≥40% stearic acid basis (GC), 4.0-5.0% Mg basis (calc on dry sub.)