Merck
  • Coupling passive sampling and time of flight mass spectrometry for a better estimation of polar pesticide freshwater contamination: Simultaneous target quantification and screening analysis.
Coupling passive sampling and time of flight mass spectrometry for a better estimation of polar pesticide freshwater contamination: Simultaneous target quantification and screening analysis.
Journal of chromatography. A (2015-02-24)
Robin Guibal, Sophie Lissalde, Adeline Charriau, Gaëlle Poulier, Nicolas Mazzella, Gilles Guibaud
ABSTRACT
The aim of this study was first to develop and validate an analytical method for the quantification of 35 polar pesticides and 9 metabolites by ultra-high-performance-liquid chromatography combined with a high resolution time-of-flight mass spectrometer detector (UHPLC-(Q)-TOF). Various analytical conditions were investigated (eluent composition and mass parameters) to optimize analyte responses. Analytical performance (linearity, limit of quantification, and accuracy) was then evaluated and interference in the extract of a passive sampler exposed in freshwater (POCIS: Polar Organic Chemical Integrative Sampler) was studied. The proposed quantification method was validated for 43 compounds with variation of calibration slopes below 10% in environmental matrix. For the unvalidated compound DIA (atrazine-desisopropyl: an atrazine metabolite), interference increased the error of concentration determination (50%). The limits of quantification obtained by combining POCIS and UHPLC-(Q)-TOF for 43 target compounds were between 0.1 (terbuthylazine) and 10.7 ng/L (acetochlor). Secondly, the method was successfully applied during a 14-day POCIS river exposure, and gave concentration values similar to a more commonly used triple quadrupole detector regarding concentration, but allowed for the detection of more compounds. Additionally with the targeted compound quantification, the (Q)-TOF mass spectrometer was also used for screening non-target compounds (other pesticides and pharmaceuticals) in POCIS extracts. Moreover, the acquisition of full scan MS data allowed the identification of the polyethylene glycol (PEG) compounds which gave unresolvable interference to DIA, and thus questions the ability of DIA to be used as performance reference compound (PRC) to determine sampling rates in situ. This study therefore illustrates the potential, and proposes a pathway, of UHPLC-(Q)-TOF combined with POCIS in situ pre-concentration for both quantitative and screening analyses of organic contaminants in water.
Sigma-Aldrich
Ammonium formate, reagent grade, 97%
Sigma-Aldrich
Ammonium formate, SAJ first grade, ≥95.0%
Supelco
Methacrifos, PESTANAL®, analytical standard
Sigma-Aldrich
Ammonium formate, ≥99.995% trace metals basis
Sigma-Aldrich
Ammonium formate, BioUltra, ≥99.0% (calc. based on dry substance, NT)
Supelco
Ammonium formate, eluent additive for LC-MS, LiChropur, ≥99.0%
Sigma-Aldrich
Ethyl acetate, suitable for HPLC
Sigma-Aldrich
Ethyl acetate, JIS 300, ≥99.5%, for residue analysis
Sigma-Aldrich
Formic acid, JIS special grade, ≥98.0%
Sigma-Aldrich
Ethyl acetate, JIS special grade, ≥99.5%
Sigma-Aldrich
Ethyl acetate, ReagentPlus®, ≥99.8%
Supelco
Isoxaben, PESTANAL®, analytical standard
Sigma-Aldrich
Formic acid, ≥95%, FCC, FG
Sigma-Aldrich
Ammonium formate solution, BioUltra, 10 M in H2O
Supelco
Ammonium formate solution, 10 mM in H2O, suitable for HPLC
Supelco
Ethyl acetate, analytical standard
Sigma-Aldrich
Ethyl acetate, ≥99%, FCC, FG
Sigma-Aldrich
Ethyl acetate, natural, ≥99%, FCC, FG
Sigma-Aldrich
Ethyl acetate, anhydrous, 99.8%
Sigma-Aldrich
Ethyl acetate, ACS reagent, ≥99.5%
Sigma-Aldrich
β-D-Allose, rare aldohexose sugar
Sigma-Aldrich
Formic acid, reagent grade, ≥95%
Sigma-Aldrich
Formic acid solution, BioUltra, 1.0 M in H2O
Sigma-Aldrich
Ethyl acetate, ACS reagent, ≥99.5%
Sigma-Aldrich
Ethyl acetate, HPLC Plus, for HPLC, GC, and residue analysis, 99.9%
Sigma-Aldrich
Ethyl acetate, suitable for HPLC, ≥99.8%
Sigma-Aldrich
Formic acid, ACS reagent, ≥96%
Sigma-Aldrich
Ethyl acetate, ACS reagent, ≥99.5%
Sigma-Aldrich
Ethyl acetate, biotech. grade, ≥99.8%
Sigma-Aldrich
Formic acid, ACS reagent, ≥88%