Merck
  • Interaction of ferrocene appended Ru(II), Rh(III) and Ir(III) dipyrrinato complexes with DNA/protein, molecular docking and antitumor activity.
Interaction of ferrocene appended Ru(II), Rh(III) and Ir(III) dipyrrinato complexes with DNA/protein, molecular docking and antitumor activity.
European journal of medicinal chemistry (2014-07-12)
Rajendra Prasad Paitandi, Rakesh Kumar Gupta, Roop Shikha Singh, Gunjan Sharma, Biplob Koch, Daya Shankar Pandey
ABSTRACT
Efficacy of the ferrocene appended piano-stool dipyrrinato complexes [(η(6)-C6H6)RuCl(fcdpm)] (1), [(η(6)-C10H14)RuCl(fcdpm)] (2), [(η(6)-C12H18)RuCl(fcdpm)] (3) [(η(5)-C5Me5)RhCl(fcdpm)] (4) and [(η(5)-C5Me5)IrCl(fcdpm)] (5) [fcdpm = 5-ferrocenyldipyrromethene] toward anticancer activity have been described. Binding of the complexes with calf thymus DNA (CT-DNA) and BSA (bovine serum albumin) have been thoroughly investigated by UV-Vis and fluorescence spectroscopy. Binding constants for 1-5 (range, 10(4)-10(5) M(-1)) validated their efficient binding with CT-DNA. Molecular docking studies revealed interaction through minor groove of the DNA, on the other hand these also interact through hydrophobic residues of the protein, particularly cavity in the subdomain IIA. In vitro anticancer activity have been scrutinized by MTT assay, acridine orange/ethidium bromide (AO/EtBr) fluorescence staining, and DNA ladder (fragmentation) assay against Dalton's Lymphoma (DL) cells. Present study revealed that rhodium complex (4) is more effective relative to ruthenium (1-3) and iridium (5) complexes.
Sigma-Aldrich
Ethidium bromide solution, 10 mg/mL
Sigma-Aldrich
1,2,3,4,5-Pentamethylcyclopentadiene, 95%
Sigma-Aldrich
1,3-Cyclohexadiene, contains 0.05% BHT as inhibitor, 97%
Sigma-Aldrich
Ruthenium(III) chloride, Ru content 45-55%
Sigma-Aldrich
Ethidium bromide solution, for fluorescence, ~1% in H2O
Sigma-Aldrich
Ethidium bromide solution, BioReagent, for molecular biology, 500 μg/mL in H2O
Sigma-Aldrich
Ethidium bromide solution, BioReagent, for molecular biology, 10 mg/mL in H2O
Sigma-Aldrich
Ethylenediaminetetraacetic acid, ≥98.0% (KT)
Sigma-Aldrich
Ethidium bromide, ≥95.0%
Sigma-Aldrich
Ethylenediaminetetraacetic acid, SAJ special grade, ≥99.0%
Sigma-Aldrich
Iridium(III) chloride, 99.8% trace metals basis
Sigma-Aldrich
Ethylenediaminetetraacetic acid disodium salt solution, BioUltra, for molecular biology, pH 8.0, ~0.5 M in H2O
Sigma-Aldrich
Ethylenediaminetetraacetic acid, BioUltra, ≥99.0% (KT)
Sigma-Aldrich
Ethidium bromide, ~95% (HPLC)
Sigma-Aldrich
Rhodium(III) chloride, 98%
Sigma-Aldrich
Ethylenediaminetetraacetic acid, 99.995% trace metals basis
Sigma-Aldrich
Iridium(III) chloride, crystalline
Sigma-Aldrich
Ferrocenecarboxaldehyde, 98%
Sigma-Aldrich
Ethylenediaminetetraacetic acid, purified grade, ≥98.5%, powder
Sigma-Aldrich
Ethylenediaminetetraacetic acid, anhydrous, crystalline, BioReagent, suitable for cell culture
Sigma-Aldrich
Ethylenediaminetetraacetic acid, BioUltra, anhydrous, ≥99% (titration)
Sigma-Aldrich
Ethylenediaminetetraacetic acid solution, 0.02% in DPBS (0.5 mM), sterile-filtered, BioReagent, suitable for cell culture
Sigma-Aldrich
Ethidium bromide, BioReagent, for molecular biology, powder
Sigma-Aldrich
Ethylenediaminetetraacetic acid, ACS reagent, 99.4-100.6%, powder