Skip to Content
Merck
All Photos(1)

Documents

214027

Sigma-Aldrich

1,2,3,4,5-Pentamethylcyclopentadiene

95%

Synonym(s):

1,2,3,4,5-Pentamethyl-1,3-cyclopentadiene

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C10H16
CAS Number:
Molecular Weight:
136.23
Beilstein:
1849832
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Quality Level

Assay

95%

refractive index

n20/D 1.474 (lit.)

bp

58 °C/13 mmHg (lit.)

density

0.87 g/mL at 25 °C (lit.)

SMILES string

CC1C(C)=C(C)C(C)=C1C

InChI

1S/C10H16/c1-6-7(2)9(4)10(5)8(6)3/h6H,1-5H3

InChI key

WQIQNKQYEUMPBM-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Mechanism of the Diels-Alder reaction of paramagneticendohedral metallofullerene and 1,2,3,4,5-pentamethylcyclopentadiene has been studied. Three-step large-scale preparation of 1,2,3,4,5-pentamethylcyclopentadiene has been reported. Reaction of 1,2,3,4,5-pentamethylcyclopentadiene (HCp*) with actinide ions in gas phase has been investigated by laser ablation mass spectrometry. It undergoes radical cation catalyzed cycloaddition with electron rich allenes to form Diels-Alder product.

Application

1,2,3,4,5-pentamethylcyclopentadiene was used as:
  • Growth modifier chemical, during metal organic chemical vapour deposition of iron from iron pentacarbonyl.
  • Ligand in "one-pot" iridium-catalyzed transformation of alcohols to amides via the intermediacy of oximes.
  • Raw material for the synthesis of [Cp*Rh(bpy)H2O]2+ (Cp* = pentamethylcyclopentadienyl, bpy = 2,2′-bipyridyl), an electron mediator in the regeneration process of NADH.

Pictograms

Flame

Signal Word

Warning

Hazard Statements

Hazard Classifications

Flam. Liq. 3

Storage Class Code

3 - Flammable liquids

WGK

WGK 3

Flash Point(F)

111.2 °F - closed cup

Flash Point(C)

44 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

The Aminium Salt and Photoinduced Electron Transfer Initiated Diels-Alder Cycloaddition of Electron-rich Allenes: Evidence for a Stepwise Mechanism and the Importance of Steric and Electronic Effects for the Reactivity of Distonic Radical Cation Intermediates.
Schmittel M, et al.
Chemistry (Weinheim An Der Bergstrasse, Germany), 2(8), 1031-1040 (1996)
Nathan A Owston et al.
Organic letters, 9(1), 73-75 (2006-12-29)
[reaction: see text] The iridium catalyst [Ir(Cp*)Cl2]2 is effective for the rearrangement of oximes to furnish amides. The reaction has been combined with catalytic transfer hydrogenation between an alcohol and alkene to allow the conversion of alcohols into amides in
Journal of Organometallic Chemistry, 472, 359-359 (1994)
An improved synthesis of 1,2,3,4,5-pentamethylcyclopentadiene.
Kohl FX and Jutzi P.
Journal of Organometallic Chemistry, 243(1), 119-121 (1983)
Synth. React. Inorg. Met.-Org. Chem. , 24, 395-395 (1994)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service