Merck
  • Bioaccessibility and digestive stability of carotenoids in cooked eggs studied using a dynamic in vitro gastrointestinal model.
Bioaccessibility and digestive stability of carotenoids in cooked eggs studied using a dynamic in vitro gastrointestinal model.
Journal of agricultural and food chemistry (2015-03-10)
Chamila Nimalaratne, Patricia Savard, Sylvie F Gauthier, Andreas Schieber, Jianping Wu
ABSTRACT
Among dietary carotenoids, lutein and zeaxanthin are known to protect against age-related macular degeneration, a leading cause of irreversible vision loss in the elderly. Egg yolk is rich in lutein and zeaxanthin, however, the effect of cooking and gastrointestinal digestion on yolk carotenoids is poorly understood. An in vitro dynamic gastrointestinal model (TIM-1) was used to investigate the digestive stability and bioaccessibility of carotenoids from boiled, fried, and scrambled eggs. Bioaccessibility but not digestive stability was significantly affected by the method of cooking. The main egg carotenoids, all-E-lutein and all-E-zeaxanthin, were stable during the digestion with average recoveries of 90 and 88%, respectively. No trans-cis isomerization of carotenoids was observed during digestion. Both all-E-lutein and all-E-zeaxanthin from scrambled eggs showed significantly lower bioaccessibility compared to boiled eggs. The results indicate that the bioaccessibility of egg carotenoids can be affected by different food preparation methods.
Sigma-Aldrich
Methanol, suitable for HPLC
Sigma-Aldrich
Methanol, JIS special grade, ≥99.8%
Sigma-Aldrich
Methanol, SAJ first grade, ≥99.5%
Sigma-Aldrich
Methanol, JIS 300, ≥99.8%, for residue analysis
Sigma-Aldrich
tert-Butyl methyl ether, SAJ special grade, ≥99.0%
Sigma-Aldrich
Methanol, NMR reference standard
Sigma-Aldrich
Methanol, suitable for HPLC, gradient grade, 99.93%
Sigma-Aldrich
Pepsin from porcine gastric mucosa, lyophilized powder, ≥3,200 units/mg protein
Sigma-Aldrich
Xanthophyll, from marigold
Sigma-Aldrich
Butylated hydroxytoluene, ≥99%, FCC, FG
Supelco
tert-Butyl methyl ether, HPLC grade, suitable for HPLC, 99.8%
Supelco
Methanol, analytical standard
Sigma-Aldrich
Methanol, anhydrous, 99.8%
Supelco
Butylated Hydroxytoluene, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
Methanol, HPLC Plus, ≥99.9%, poly-coated bottles
Sigma-Aldrich
Methanol, SAJ special grade
Sigma-Aldrich
Methanol, Laboratory Reagent, ≥99.6%
Sigma-Aldrich
Methanol, ACS spectrophotometric grade, ≥99.9%
Sigma-Aldrich
tert-Butyl methyl ether, puriss. p.a., ≥99.5% (GC)
Sigma-Aldrich
Methanol, ACS reagent, ≥99.8%
Sigma-Aldrich
tert-Butyl methyl ether, ACS reagent, ≥99.0%
Sigma-Aldrich
Methanol, BioReagent, ≥99.93%
Sigma-Aldrich
Methanol, ACS reagent, ≥99.8%
Sigma-Aldrich
tert-Butyl methyl ether, reagent grade, 98%
Sigma-Aldrich
Methanol, puriss., meets analytical specification of Ph Eur, ≥99.7% (GC)
Sigma-Aldrich
Methanol, ACS reagent, ≥99.8%
Supelco
Methanol, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
Methanol, HPLC Plus, ≥99.9%
Sigma-Aldrich
Methanol, suitable for HPLC, ≥99.9%
Sigma-Aldrich
Methanol, suitable for HPLC, gradient grade, suitable as ACS-grade LC reagent, ≥99.9%