Skip to Content
Merck
All Photos(1)

Documents

X6250

Sigma-Aldrich

Xanthophyll

from marigold

Synonym(s):

α-Carotene-3,3′-diol, β, ε-carotene, β, ε-carotene-3,3′-diol, Lutein

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C40H56O2
CAS Number:
Molecular Weight:
568.87
Beilstein:
2068547
EC Number:
MDL number:
UNSPSC Code:
12352205
PubChem Substance ID:
NACRES:
NA.79

biological source

marigold

Quality Level

form

powder

color

orange to brown

shipped in

dry ice

storage temp.

−70°C

SMILES string

CC(=C/C=C/C=C(C)/C=C/C=C(C)/C=C/C1=C(C)C[C@@H](O)CC1(C)C)\C=C\C=C(C)\C=C\[C@H]2C(C)=C[C@H](O)CC2(C)C

InChI

1S/C40H56O2/c1-29(17-13-19-31(3)21-23-37-33(5)25-35(41)27-39(37,7)8)15-11-12-16-30(2)18-14-20-32(4)22-24-38-34(6)26-36(42)28-40(38,9)10/h11-25,35-37,41-42H,26-28H2,1-10H3/b12-11+,17-13+,18-14+,23-21+,24-22+,29-15+,30-16+,31-19+,32-20+/t35-,36+,37-/m0/s1

InChI key

KBPHJBAIARWVSC-RGZFRNHPSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Xanthophylls are oxygenated derivatives of carotenoids. It consists of oxygen atoms. Xanthophylls are yellow pigments and are found majorly in the leaves of most green plants.

Application

Xanthophyll has been used:
  • to quantify circulating lutein in birds
  • to study its effect on the synthesis of factor D (FD) by adipocytes
  • for the quantification of carotenoids from the leaves of Brassica oleracea

Biochem/physiol Actions

Xanthophyll/Lutein functions as an accessory light-harvesting pigment. It also acts as quenchers of singlet oxygen and chlorophyll triplet states to exhibit protection against photooxidative damage. Xanthophyll is involved in photosynthesis. It has antioxidant properties.
Dietary carotenoid with no vitamin A potency. Increases macular pigment concentration in the eye and may improve visual function.

Packaging

Dry powder packed under Argon.

Quality

Minimum 70% total carotenoids as xanthophyll.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Effect of green juice and their bioactive compounds on genotoxicity induced by alkylating agents in mice
Fagundes GE, et al.
Journal of Toxicology and Environmental Health. Part A, 80(13-15), 756-766 (2017)
Lutein leads to a decrease of factor D secretion by cultured mature human adipocytes
Tian Y, et al.
Journal of Ophthalmology, 2015 (2015)
Characterisation of Carotenoids Involved in the Xanthophyll Cycle
Kuczynska P, et al.
Carotenoids, 1-1 (2017)
Extraction and purification of carotenoids from vegetables
Rebecca LJ, et al.
Journal of Chemical and Pharmaceutical Research, 6(4), 594-598 (2014)
Carotenoids, birdsong and oxidative status: administration of dietary lutein is associated with an increase in song rate and circulating antioxidants (albumin and cholesterol) and a decrease in oxidative damage
Casagrande S, et al.
PLoS ONE, 9(12), e115899-e115899 (2014)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service