Merck
  • CYP-independent inhibition of platelet aggregation in rabbits by a mixed disulfide conjugate of clopidogrel.
CYP-independent inhibition of platelet aggregation in rabbits by a mixed disulfide conjugate of clopidogrel.
Thrombosis and haemostasis (2014-09-19)
H Zhang, D A Lauver, P F Hollenberg
ABSTRACT
Dual antiplatelet therapy with clopidogrel and aspirin has been the standard of care in the United States for patients with acute coronary syndromes (ACS) and/or undergoing percutaneous coronary interventions (PCI). However, the effectiveness of clopidogrel varies significantly among different sub-populations due to inter-individual variability. In this study we examined the antiplatelet potential of a novel mixed disulfide conjugate of clopidogrel with the aim to overcome the inter-individual variability. In the metabolic studies using human liver microsomes and cDNA-expressed P450s, we confirmed that multiple P450s are involved in the bioactivation of 2-oxoclopidogrel to H4, one of the diastereomers of the pharmacologically active metabolite (AM) possessing antiplatelet activity. Results from kinetic studies demonstrated that 2C19 is the most active in converting 2-oxoclopidogrel to H4 with a catalytic efficiency of 0.027 µM⁻¹min⁻¹ in the reconstituted system. On the basis of this finding, we were able to biosynthesise the conjugate of clopidogrel with 3-nitropyridine-2-thiol, referred to as clopNPT, and examined its antiplatelet activity in male New Zealand white rabbits. After administration as intravenous bolus at 2 mg/kg, the clopNPT conjugate was rapidly converted to the AM leading to the inhibition of platelet aggregation (IPA). Analyses of the blood samples drawn at various time points showed that intravenous administration of clopNPT led to ~70% IPA within 1 hour and the IPA persisted for more than 3 hours. Since the antiplatelet activity of clopNPT does not require bioactivation by P450s, the mixed disulfide conjugate of clopidogrel has the potential to overcome the inter-individual variability in clopidogrel therapy.
Sigma-Aldrich
Citrate Concentrated Solution, BioUltra, for molecular biology, 1 M in H2O
Sigma-Aldrich
Citrate Concentrated Solution, BioReagent, suitable for coagulation assays, 4 % (w/v)
Sigma-Aldrich
3-Nitropyridine-2-thiol, 96%
Supelco
Meprobamate solution, 1.0 mg/mL in methanol, ampule of 1 mL, certified reference material, Cerilliant®
Lysine hydrochloride, European Pharmacopoeia (EP) Reference Standard
Supelco
Sodium Citrate, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
2-Bromo-3′-methoxyacetophenone, 98%
Sigma-Aldrich
Formic acid solution, BioUltra, 1.0 M in H2O
Supelco
L-Lysine hydrochloride solution, 100 mM amino acid in 0.1 M HCl, analytical standard
Sigma-Aldrich
L-Lysine monohydrochloride, BioUltra, ≥99.5% (AT)
Sigma-Aldrich
Formic acid, ≥95%, FCC, FG
Sigma-Aldrich
5-(N,N-Dimethyl)amiloride hydrochloride
Supelco
Meprobamate, analytical standard
Sigma-Aldrich
L-Lysine monohydrochloride, reagent grade, ≥98% (HPLC)
Sigma-Aldrich
L-Lysine monohydrochloride, from non-animal source, meets EP, JP, USP testing specifications, suitable for cell culture, 98.5-101.0%
Sigma-Aldrich
L-Lysine monohydrochloride, SAJ special grade, ≥99.0%
Sigma-Aldrich
Formic acid, JIS special grade, ≥98.0%
Supelco
L-Lysine monohydrochloride, certified reference material, TraceCERT®, Manufactured by: Sigma-Aldrich Production GmbH, Switzerland
Supelco
L-Lysine monohydrochloride, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
Formic acid, ACS reagent, ≥96%
Sigma-Aldrich
Formic acid, reagent grade, ≥95%
Sigma-Aldrich
Formic acid, puriss., meets analytical specifications of DAC, FCC, 98.0-100%
Sigma-Aldrich
Formic acid, puriss. p.a., ACS reagent, reag. Ph. Eur., ≥98%
Sigma-Aldrich
Formic acid, ACS reagent, ≥88%