Merck
  • Viola mandshurica ethanolic extract prevents high-fat-diet-induced obesity in mice by activating AMP-activated protein kinase.
Viola mandshurica ethanolic extract prevents high-fat-diet-induced obesity in mice by activating AMP-activated protein kinase.
Environmental toxicology and pharmacology (2014-06-01)
Yoon-Young Sung, Dong-Seon Kim, Ho Kyoung Kim
ABSTRACT
Viola mandshurica W. Becker has been used as an expectorant, diuretic, and anti-inflammatory agent. We evaluated the effects of V. mandshurica ethanol extract (VME) on high-fat-diet (HFD)-induced obesity in mice. HPLC analysis showed that the VME contained 11.95 ± 0.37 mg/g esculetin and 0.13 ± 0.01 mg/g scopoletin. Orally administered VME decreased the body weight, adipose tissue mass, adipocyte size, and triglyceride and leptin serum concentrations. In contrast, VME increased serum adiponectin concentrations and adiponectin expression levels in epididymal adipose tissues. VME also significantly reversed the HFD-induced elevation of the mRNA and protein levels of lipogenic genes such as peroxisome proliferator-activated receptor γ, sterol regulatory element-binding protein-1c, fatty-acid synthase, and adipocyte protein 2. Moreover, VME reversed the HFD-induced inhibition of AMP-activated protein kinase (AMPK) and acetyl-coA carboxylase phosphorylation in epididymal adipose tissues. Furthermore, treatment of VME and esculetin in 3T3-L1 cells inhibited adipocyte differentiation and fat accumulation. These results suggest that VME exerts anti-obesity effects in HFD-induced obese mice by activating AMPK and suppressing PPARγ expression in adipose tissues.
Sigma-Aldrich
Acetonitrile solution, contains 0.05 % (v/v) trifluoroacetic acid
Sigma-Aldrich
Acetonitrile solution, contains 0.1 % (v/v) trifluoroacetic acid, suitable for HPLC
Sigma-Aldrich
Acetonitrile solution, contains 0.1 % (v/v) formic acid, suitable for HPLC
Sigma-Aldrich
Acetonitrile solution, contains 10.0% acetone, 0.05% formic acid, 40.0% 2-propanol
Sigma-Aldrich
Acetonitrile solution, contains 0.05 % (w/v) ammonium formate, 5 % (v/v) water, 0.1 % (v/v) formic acid, suitable for HPLC
Supelco
Residual Solvent - Acetonitrile, Pharmaceutical Secondary Standard; Certified Reference Material
Supelco
Acetonitrile, analytical standard
Sigma-Aldrich
Acetonitrile, anhydrous, 99.8%
Sigma-Aldrich
Acetonitrile, ≥99.5%, ACS reagent
Sigma-Aldrich
6,7-Dihydroxycoumarin, 98%
Supelco
Acetonitrile, HPLC grade, ≥99.93%
Sigma-Aldrich
Scopoletin, ≥99%
Supelco
Scopoletin, analytical standard
Sigma-Aldrich
Acetonitrile, for chromatography
Sigma-Aldrich
Acetonitrile, electronic grade, 99.999% trace metals basis
Sigma-Aldrich
Acetonitrile, for residue analysis, JIS 5000
Sigma-Aldrich
Acetonitrile, HPLC Plus, ≥99.9%, poly-coated bottles
Sigma-Aldrich
Acetonitrile, ≥99.8%, for residue analysis, JIS 300
Sigma-Aldrich
Acetonitrile, ≥99.8%, for residue analysis, JIS 1000
Sigma-Aldrich
Acetonitrile, JIS special grade, ≥99.5%
Sigma-Aldrich
Acetonitrile, ≥99.8%, suitable for HPLC
Sigma-Aldrich
Acetonitrile, SAJ first grade, ≥99.0%
USP
Residual Solvent Class 2 - Acetonitrile, United States Pharmacopeia (USP) Reference Standard
Sigma-Aldrich
Acetonitrile, ReagentPlus®, 99%
Esculetin, European Pharmacopoeia (EP) Reference Standard
Supelco
Acetonitrile, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
Acetonitrile, biotech. grade, ≥99.93%
Sigma-Aldrich
Acetonitrile, suitable for DNA synthesis, ≥99.9% (GC)
Sigma-Aldrich
Acetonitrile, ACS reagent, ≥99.5%
Sigma-Aldrich
Acetonitrile, suitable for HPLC, gradient grade, ≥99.9%