Merck
  • Potential dangers from topical preparations containing methyl salicylate.
Potential dangers from topical preparations containing methyl salicylate.
Human & experimental toxicology (1996-09-01)
T Y Chan
ABSTRACT
Methyl salicylate (oil of wintergreen) is widely available in many over-the-counter liniments, ointments, lotions or medicated oils for the relief of musculoskeletal aches and pains. Ingestion of methyl salicylate poses the threat of severe, rapid-onset salicylate poisoning because of its liquid, concentrated form and lipid solubility. Excessive usage of these preparations in patients receiving warfarin may result in adverse interactions and bleedings. Methyl salicylate in topical analgesic preparations may cause irritant or allergic contact dermatitis and anaphylactic reactions. Physicians should fully appreciate the potential dangers from topical preparations containing methyl salicylate.
Sigma-Aldrich
Methyl salicylate, SAJ first grade, ≥98.0%
Sigma-Aldrich
Methyl salicylate, ReagentPlus®, ≥99% (GC)
USP
Methyl salicylate, United States Pharmacopeia (USP) Reference Standard
Supelco
Methyl salicylate, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
Methyl salicylate, ≥98%, FCC, FG
Supelco
Methyl salicylate, analytical standard
Sigma-Aldrich
Methyl salicylate, tested according to Ph. Eur.