Merck
  • [Occupational exposure-associated hematological and immunologic manifestations to the benzene-toluene-xylene (Btx) mixture].
[Occupational exposure-associated hematological and immunologic manifestations to the benzene-toluene-xylene (Btx) mixture].
Revista medica del Instituto Mexicano del Seguro Social (2009-03-07)
Luis Cuauhtémoc Haro-García, César Raúl González-Bonilla, Rommel Chacón-Salinas, Carlos Pérez-Lucio, Cuauhtémoc Arturo Juárez-Pérez, Víctor Hugo Borja-Aburto
ABSTRACT
Despite, the idea promoted to study occupational exposure to benzene and its mixture with toluene and xylene (BTX) because it appears to determine its toxicity and probably the production of additive effects, it persists interest to recognizing its hematological and immunotoxic effects. The fact that exposure to a sole substance in the occupational field is infrequent. Available contributions that analyze these implications are scarce, with contradictory results, and in their majority are limited to the fraction of benzene. Epidemiologic studies that have evaluated occupational exposure to any of the BTX fractions have been based on personal monitoring, while others have characterized this heterogeneously and are accompanied by weaker proposals. The conformation of specific methods to stimulate occupational exposure to the BTX mixture would contribute to its homogenization and allow for a more integral view in terms of determining BTX exposure. On the other hand, the application of BTX exposure biomarkers has been questioned in studies contemplating the specific biological effects of reference-associated chronic exposure. Analysis of the hematological and immunologic manifestations associated BTX mixture is based on information that is unclear, controversial, or even speculative to date.
Product Description
Supelco
Toluene solution, certified reference material, 5000 μg/mL in methanol
Supelco
Toluene solution, certified reference material, TraceCERT®, 200 μg/mL in methanol, ampule of 1 mL
Supelco
Residual Solvent - Toluene, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
Toluene, JIS 1000, suitable for residue analysis, ≥99.8%
Sigma-Aldrich
Toluene, SAJ first grade, ≥99.0%
Supelco
Toluene, HPLC grade, 99.8%
Supelco
Toluene, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
Toluene, puriss. p.a., ACS reagent, reag. ISO, reag. Ph. Eur., ≥99.7% (GC)
Sigma-Aldrich
Toluene, suitable for HPLC, 99.9%
Sigma-Aldrich
Toluene, ACS reagent, ≥99.5%
Sigma-Aldrich
Toluene, ACS reagent, ≥99.5%
Sigma-Aldrich
Toluene, ACS reagent, ≥99.5%
Sigma-Aldrich
Toluene, anhydrous, 99.8%
Sigma-Aldrich
Toluene, suitable for HPLC, ≥99.9%
Sigma-Aldrich
Toluene, Laboratory Reagent, ≥99.3%
Sigma-Aldrich
Toluene, HPLC Plus, for HPLC, GC, and residue analysis, ≥99.9%
Supelco
Toluene, analytical standard
Sigma-Aldrich
Toluene, JIS special grade, ≥99.5%
Sigma-Aldrich
Toluene, JIS 300, suitable for residue analysis, ≥99.8%