Merck
Fragrance material review on methyl salicylate.
Food and chemical toxicology : an international journal published for the British Industrial Biological Research Association (2007-11-23)
A Lapczynski, L Jones, D McGinty, S P Bhatia, C S Letizia, A M Api
ABSTRACT
A toxicologic and dermatologic review of methyl salicylate when used as a fragrance ingredient is presented.
Sigma-Aldrich
Methyl salicylate, ReagentPlus®, ≥99% (GC)
USP
Methyl salicylate, United States Pharmacopeia (USP) Reference Standard
Sigma-Aldrich
Methyl salicylate, SAJ first grade, ≥98.0%
Supelco
Methyl salicylate, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
Methyl salicylate, ≥98%, FCC, FG
Supelco
Methyl salicylate, analytical standard
Sigma-Aldrich
Methyl salicylate, tested according to Ph. Eur.