Merck
Renal papillary necrosis induced by naproxen.
Pediatric nephrology (Berlin, Germany) (2003-05-30)
Larisa Kovacevic, Jay Bernstein, Rudolph P Valentini, Abubakr Imam, Neena Gupta, Tej K Mattoo
ABSTRACT
A 17-year-old healthy girl was admitted to our hospital with diffuse abdominal pain and decreased oral intake of about 11 days duration. About a week prior to admission, she had taken naproxen, 250 mg four times a day for 4 days. Physical examination was normal except for diffuse abdominal tenderness on deep palpation. Investigations revealed high serum BUN (42 mg/dl) and creatinine (4.0 mg/dl). Serum electrolytes and complement (C3, C4) levels and urinalysis were normal. Antinuclear-antibody and anti-dsDNA were negative. Kidney biopsy revealed renal papillary necrosis, acute tubular necrosis, and focal interstitial nephritis. A diagnosis of nonoliguric acute renal failure due to naproxen nephrotoxicity was made. She received intravenous hydration, and oral steroids, which was gradually discontinued in 3 months. A follow-up at 4 months revealed normal renal function with a serum creatinine of 1.1 mg/dl, BUN 7 mg/dl, and normal urinalysis. The report highlights a need for caution while using naproxen or any other nonsteroidal anti-inflammatory drugs, even for a short duration.
Product Description
Supelco
Naproxen, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
Naproxen, meets USP testing specifications
Sigma-Aldrich
(S)-(+)-6-Methoxy-α-methyl-2-naphthaleneacetic acid, 98%
Supelco
Naproxen solution, 1.0 mg/mL in methanol, ampule of 1 mL, certified reference material, Cerilliant®
Naproxen, European Pharmacopoeia (EP) Reference Standard
Supelco
Naproxen, VETRANAL®, analytical standard
USP
Naproxen, United States Pharmacopeia (USP) Reference Standard