Merck
  • Explorations of substituted urea functionality for the discovery of new activators of the heme-regulated inhibitor kinase.
Explorations of substituted urea functionality for the discovery of new activators of the heme-regulated inhibitor kinase.
Journal of medicinal chemistry (2013-11-23)
Ting Chen, Khuloud Takrouri, Sung Hee-Hwang, Sandeep Rana, Revital Yefidoff-Freedman, Jose Halperin, Amarnath Natarajan, Christophe Morisseau, Bruce Hammock, Michael Chorev, Bertal H Aktas
ABSTRACT
Heme-regulated inhibitor kinase (HRI), a eukaryotic translation initiation factor 2 alpha (eIF2α) kinase, plays critical roles in cell proliferation, differentiation, and adaptation to cytoplasmic stress. HRI is also a critical modifier of hemoglobin disorders such as β-thalassemia. We previously identified N,N'-diarylureas as potent activators of HRI suitable for studying the biology of this important kinase. To expand the repertoire of chemotypes that activate HRI, we screened a ∼1900 member N,N'-disubstituted urea library in the surrogate eIF2α phosphorylation assay, identifying N-aryl,N'-cyclohexylphenoxyurea as a promising scaffold. We validated hit compounds as a bona fide HRI activators in secondary assays and explored the contributions of substitutions on the N-aryl and N'-cyclohexylphenoxy groups to their activity by studying focused libraries of complementing analogues. We tested these N-aryl,N'-cyclohexylphenoxyureas in the surrogate eIF2α phosphorylation and cell proliferation assays, demonstrating significantly improved bioactivities and specificities. We consider these compounds to represent lead candidates for the development of potent and specific HRI activators.
Sigma-Aldrich
Methyl 2-isocyanatobenzoate, 97%
Urea, European Pharmacopoeia (EP) Reference Standard
Sigma-Aldrich
Urea, BioUltra, for molecular biology, 99% (T)
Sigma-Aldrich
Urea, powder, BioReagent, for molecular biology, suitable for cell culture
Sigma-Aldrich
Urea, suitable for electrophoresis
Sigma-Aldrich
Urea, ReagentPlus®, ≥99.5%, pellets
Sigma-Aldrich
Urea, puriss. p.a., ACS reagent, reag. Ph. Eur., ≥99%
Sigma-Aldrich
Urea, meets USP testing specifications
Supelco
Urea, 8 M (after reconstitution with 16 mL high purity water)
Sigma-Aldrich
Urea, ACS reagent, 99.0-100.5%
Sigma-Aldrich
Urea, puriss., meets analytical specification of Ph. Eur., BP, USP, 99.0-100.5%, 99.0-101.0% (calc. on dry substance)
Sigma-Aldrich
Urea, BioXtra, pH 7.5-9.5 (20 °C, 5 M in H2O)
Sigma-Aldrich
Urea, JIS special grade, ≥99.0%
Sigma-Aldrich
Urea, ≥99.0%
Sigma-Aldrich
Urea, SAJ first grade, ≥98.0%
Millipore
Urea solution, suitable for microbiology, 40% in H2O
Sigma-Aldrich
Urea-12C, 99.9 atom % 12C
Sigma-Aldrich
Urea solution, 40 % (w/v) in H2O
Sigma-Aldrich
Urea solution, BioUltra, ~8 M in H2O