Merck
  • An ectotherm homologue of human predicted gene NAT16 encodes histidine N-acetyltransferase responsible for Nα-acetylhistidine synthesis.
An ectotherm homologue of human predicted gene NAT16 encodes histidine N-acetyltransferase responsible for Nα-acetylhistidine synthesis.
Biochimica et biophysica acta (2013-10-15)
Shoji Yamada, Shiori Arikawa
ABSTRACT
Nα-Acetylhistidine (NAH) is present in very high concentrations exclusively in the brain and lens of ectothermic vertebrates, including ray-finned fishes, amphibians and reptiles, and not in those of endothermic birds and mammals. Although NAH is known to be synthesized from l-His and acetyl-CoA by histidine N-acetyltransferase (HISAT; EC 2.3.1.33), the gene encoding HISAT has remained unknown for any organism. HISAT was purified from the blue mackerel brain, and its partial amino acid sequences were analyzed using mass spectrometry and Edman degradation. Using the sequence information, the corresponding gene was cloned and sequenced. Recombinant proteins encoded by the fish gene and its human homologue were expressed in a cell-free translation system. HISAT was identified to be a protein encoded by a fish homologue of the human predicted gene NAT16 (N-acetyltransferase 16). HISAT is an unstable enzyme that is rapidly and irreversibly inactivated during preincubation at 37°C in the absence of acetyl-CoA. In fish brain, the HISAT gene is expressed as two splice variants containing an identical ORF but differing lengths of 5'-UTR. Both variants are expressed exclusively in the fish brain and lens. Interestingly, the recombinant human NAT16 protein, unlike the recombinant fish HISAT, has only trace enzyme activity for NAH synthesis. These results propose that the function of mammalian NAT16 has been altered from l-His acetylation (NAH synthesis) to another different biological role. The molecular identification of HISAT will allow progress in the understanding of the physiological function of NAH in ectothermic vertebrates.
Sigma-Aldrich
Agarose, High EEO
Sigma-Aldrich
Agarose, Low EEO, for Immunoelectrophoresis
Sigma-Aldrich
Agarose, Special High EEO
Sigma-Aldrich
Agarose, Low EEO
Sigma-Aldrich
Agarose, Wide range, for molecular biology
Sigma-Aldrich
Agarose, High EEO, for molecular biology
Sigma-Aldrich
L-Histidine, BioUltra, ≥99.5% (NT)
Sigma-Aldrich
Agarose, BioReagent, for molecular biology, Wide range/Standard 3:1
Sigma-Aldrich
Agarose, Medium EEO, for molecular biology
Sigma-Aldrich
Agarose, High Gelling Temperature
Sigma-Aldrich
Agarose, Ultra-low Gelling Temperature, molecular biology grade
Sigma-Aldrich
L-Histidine, suitable for cell culture, meets EP, USP testing specifications, from non-animal source
Sigma-Aldrich
L-Histidine, ReagentPlus®, ≥99% (TLC)
Sigma-Aldrich
Sepharose 6B, 6% Beaded Agarose, 45-165 μm (wet), fractionation range 10,000-1,000,000 Da (dextrans)
Sigma-Aldrich
Agarose, Type I, low EEO
Sigma-Aldrich
Agarose, BioReagent, for molecular biology, low EEO
Sigma-Aldrich
Agarose, For pulsed field electrophoresis running gel
Sigma-Aldrich
Agarose, Type I-A, low EEO
Supelco
L-Histidine, certified reference material, TraceCERT®, Manufactured by: Sigma-Aldrich Production GmbH, Switzerland
Supelco
L-Histidine, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
Agarose, Ultra-low Gelling Temperature
Sigma-Aldrich
Agarose, Type II-A, Medium EEO
Sigma-Aldrich
Agarose, Type IV, Special High EEO
Sigma-Aldrich
Agarose, for molecular biology
SAFC
L-Histidine
Sigma-Aldrich
Sepharose 4B, 45-165 μm bead diameter