Merck
  • Combined losartan and nitro-oleic acid remarkably improves diabetic nephropathy in mice.
Combined losartan and nitro-oleic acid remarkably improves diabetic nephropathy in mice.
American journal of physiology. Renal physiology (2013-08-16)
Ying Liu, Zhanjun Jia, Shanshan Liu, Maicy Downton, Gang Liu, Yaomin Du, Tianxin Yang
ABSTRACT
Diabetic nephropathy (DN) is a leading cause of end-stage renal disease (ESRD). The inhibitors of renin-angiotensin-aldosterone system (RAAS) can alleviate some of the symptoms of DN but fail to stop the progression to ESRD. Our previous studies demonstrate renoprotective action of nitro-oleic acid (OA-NO2) in several rodent models of renal disease. Here we examined the therapeutic potential and the underlying mechanism of combination of losartan and OA-NO2 in db/db mice. OA-NO2 was infused at 5 mg·kg(-1)·day(-1) via osmotic minipump, and losartan was incorporated into diet at 10 mg·kg(-1)·day(-1), each administered alone or in combination for 2 wk. Diabetic db/db mice developed progressive albuminuria and glomerulosclerosis, accompanied by podocytes loss, increased indexes of renal fibrosis, oxidative stress, and inflammation. Treatment of the diabetic mice with OA-NO2 or losartan alone moderately ameliorated kidney injury; however, the combined treatment remarkably reduced albuminuria, restored glomerular filtration barrier structure, and attenuated glomerulosclerosis, accompanied with significant suppression of renal oxidative stress and inflammation. These data demonstrate that combination of losartan and OA-NO2 effectively reverses renal injury in DN.
Sigma-Aldrich
Oleic acid, SAJ first grade, ≥70.0%
Sigma-Aldrich
Oleic acid, technical grade, 90%
Supelco
Oleic acid, analytical standard
Sigma-Aldrich
Oleic acid, ≥99% (GC)
Sigma-Aldrich
Oleic acid, BioReagent, suitable for cell culture
Sigma-Aldrich
Oleic acid, meets analytical specification of Ph, Eur., 65.0-88.0% (GC)
Sigma-Aldrich
Oleic acid, natural, FCC
Sigma-Aldrich
Anti-β-Actin antibody, Mouse monoclonal, clone AC-15, purified from hybridoma cell culture
Supelco
Oleic acid, Selectophore, ≥99%
Supelco
Losartan potassium
Sigma-Aldrich
Anti-Fibronectin antibody produced in rabbit, affinity isolated antibody, buffered aqueous solution
Losartan potassium, European Pharmacopoeia (EP) Reference Standard