Merck
  • Long-term coffee consumption in relation to fracture risk and bone mineral density in women.
Long-term coffee consumption in relation to fracture risk and bone mineral density in women.
American journal of epidemiology (2013-07-25)
Helena Hallström, Liisa Byberg, Anders Glynn, Eva Warensjö Lemming, Alicja Wolk, Karl Michaëlsson
ABSTRACT
High consumption of coffee has been suggested to reduce the risk of some late-onset diseases and death but also to contribute to the development of osteoporotic fractures. Results of previous fracture studies have been inconsistent, and a comprehensive study is needed. The longitudinal population-based Swedish Mammography Cohort, including 61,433 women born in 1914-1948, was followed up from 1987 through 2008. Coffee consumption was assessed with repeated food frequency questionnaires. During follow-up, 14,738 women experienced fracture of any type, and 3,871 had a hip fracture. In a subcohort (n = 5,022), bone density was measured and osteoporosis determined (n = 1,012). After multivariable adjustment, there was no evidence of a higher rate of any fracture (hazard ratio per 200 mL coffee = 0.99; 95% confidence interval: 0.98, 1.00) or hip fracture (hazard ratio per 200 mL coffee = 0.97, 95% confidence interval: 0.95, 1.00) with increasing coffee consumption. A high coffee intake (≥4 cups daily) versus a low intake (<1 cup daily) was associated with a 2%-4% lower bone density, depending on site (P < 0.001), but the odds ratio for osteoporosis was only 1.28 (95% confidence interval: 0.88, 1.87). Thus, high coffee consumption was associated with a small reduction in bone density that did not translate into an increased risk of fracture.
Product Description
Supelco
Melting point standard 235-237°C, analytical standard
Supelco
Caffeine solution, analytical standard, 1.0 mg/mL in methanol
Supelco
Caffeine solution, 1.0 mg/mL in methanol, ampule of 1 mL, certified reference material, Cerilliant®
Caffeine for system suitability, European Pharmacopoeia (EP) Reference Standard
Sigma-Aldrich
Caffeine, anhydrous, tested according to Ph. Eur.
Supelco
Mettler-Toledo Calibration substance ME 18872, Caffeine, traceable to primary standards (LGC)
Sigma-Aldrich
Caffeine, anhydrous, 99%, FCC, FG
Supelco
Caffeine, Pharmaceutical Secondary Standard; Certified Reference Material
Supelco
Caffeine Melting Point Standard, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
Caffeine, SAJ special grade, ≥98.5%
Sigma-Aldrich
Caffeine, Sigma Reference Standard, vial of 250 mg
Sigma-Aldrich
Caffeine, BioXtra
Sigma-Aldrich
Caffeine, meets USP testing specifications, anhydrous
Sigma-Aldrich
Caffeine, powder, ReagentPlus®
Supelco
Caffeine, certified reference material, TraceCERT®, Manufactured by: Sigma-Aldrich Production GmbH, Switzerland
Caffeine, European Pharmacopoeia (EP) Reference Standard