Skip to Content
Merck
All Photos(2)

Key Documents

C8960

Sigma-Aldrich

Caffeine

BioXtra

Synonym(s):

1,3,7-Trimethylxanthine

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C8H10N4O2
CAS Number:
Molecular Weight:
194.19
Beilstein:
17705
EC Number:
MDL number:
UNSPSC Code:
12352210
PubChem Substance ID:
NACRES:
NA.31
Pricing and availability is not currently available.

product line

BioXtra

Quality Level

form

powder

impurities

≤0.0005% Phosphorus (P)
≤0.1% Insoluble matter

ign. residue

≤0.1%

mp

234-236.5 °C (lit.)

solubility

chloroform: 0.5 M, clear, colorless

anion traces

chloride (Cl-): ≤0.05%
sulfate (SO42-): ≤0.05%

cation traces

Al: ≤0.0005%
Ca: ≤0.001%
Cu: ≤0.0005%
Fe: ≤0.0005%
K: ≤0.005%
Mg: ≤0.001%
NH4+: ≤0.05%
Na: ≤0.005%
Pb: ≤0.001%
Zn: ≤0.0005%

SMILES string

CN1C(=O)N(C)c2ncn(C)c2C1=O

InChI

1S/C8H10N4O2/c1-10-4-9-6-5(10)7(13)12(3)8(14)11(6)2/h4H,1-3H3

InChI key

RYYVLZVUVIJVGH-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Precautionary Statements

Hazard Classifications

Acute Tox. 4 Oral

Storage Class Code

11 - Combustible Solids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

CONSORT extension for reporting N-of-1 trials (CENT) 2015: Explanation and elaboration.
Larissa Shamseer et al.
BMJ (Clinical research ed.), 350, h1793-h1793 (2015-05-16)
Nicolas Bousette et al.
Journal of cellular physiology, 229(3), 374-383 (2013-09-17)
Calnexin (CNX) is an endoplasmic reticulum (ER) quality control chaperone that has been implicated in ER stress. ER stress is a prominent pathological feature of various pathologic conditions, including cardiovascular diseases. However, the role of CNX and ER stress has
Louise A Horrigan et al.
Pharmacology & therapeutics, 111(3), 877-892 (2006-03-17)
Caffeine is a member of the methylxanthine family of drugs, and is the most widely consumed behaviourally active substance in the western world. This article is focused on the impact of caffeine on immune function. In this regard, a number
Jonathan M Hodgson et al.
The American journal of clinical nutrition, 97(5), 943-950 (2013-04-05)
Measures of blood pressure variation have been associated with cardiovascular disease and related outcomes. The regular consumption of black tea can lower blood pressure, but its effects on blood pressure variation have yet to be investigated. We aimed to assess
Rohit A Sinha et al.
Hepatology (Baltimore, Md.), 59(4), 1366-1380 (2013-08-10)
Caffeine is one of the world's most consumed drugs. Recently, several studies showed that its consumption is associated with lower risk for nonalcoholic fatty liver disease (NAFLD), an obesity-related condition that recently has become the major cause of liver disease

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service