Merck
  • Antibiotic prophylaxis for hysterectomy, a prospective cohort study: cefuroxime, metronidazole, or both?
Antibiotic prophylaxis for hysterectomy, a prospective cohort study: cefuroxime, metronidazole, or both?
BJOG : an international journal of obstetrics and gynaecology (2013-06-22)
T H I Brummer, A-M Heikkinen, J Jalkanen, J Fraser, J Mäkinen, E Tomás, T Seppälä, J Sjöberg, P Härkki
ABSTRACT
To evaluate cefuroxime and metronidazole antibiotic prophylaxis. Observational nonrandomised 1-year prospective cohort study. Fifty-three hospitals in Finland. A total of 5279 women undergoing hysterectomy for benign indications, with cefuroxime given to 4301 and metronidazole given to 2855. Excluding other antibiotics, cefuroxime alone was given to 2019, metronidazole alone was given to 518, and they were administered in combination to 2252 women. Data on 1115 abdominal hysterectomies (AHs), 1541 laparoscopic hysterectomies (LHs), and 2133 vaginal hysterectomies (VHs) were analysed using logistic regression adjusted for confounding factors. Postoperative infections. Cefuroxime had a risk-reductive effect for total infections (adjusted odds ratio, OR, 0.29; 95% confidence interval, 95% CI, 0.22-0.39), but the independent effect of metronidazole and the interaction effect of cefuroxime and metronidazole were nonsignificant. In subgroup analyses of AHs, LHs, and VHs involving those receiving the two main antibiotics only, the effect of cefuroxime alone nonsignificantly differed from that of cefuroxime and metronidazole in combination for all types of infection. The absence of cefuroxime, assessed by comparing metronidazole alone with cefuroxime and metronidazole in combination, led to an increased risk for total infections in AHs (adjusted OR 3.63; 95% CI 1.99-6.65), in LHs (OR 3.53; 95% CI 1.74-7.18), and in VHs (OR 4.05; 95% CI 2.30-7.13), and also increased risks for febrile events in all categories (AHs, OR 2.86; 95% CI 1.09-7.46; LHs, OR 13.19; 95% CI 3.66-47.49; VHs, OR 12.74; 95% CI 3.01-53.95), wound infections in AHs (OR 6.88; 95% CI 1.09-7.49), and pelvic infections in VHs (OR 4.26; 95% CI 1.76-10.31). In this study, cefuroxime appeared to be effective in prophylaxis against infections. Metronidazole appeared to be ineffective, with no additional risk-reductive effect when combined with cefuroxime.
Product Description
Supelco
Metronidazole, VETRANAL®, analytical standard
Supelco
Metronidazole, analytical standard
Sigma-Aldrich
Metronidazole, BioXtra
Sigma-Aldrich
Metronidazole, SAJ first grade, ≥99.0%
Supelco
Metronidazole, Pharmaceutical Secondary Standard; Certified Reference Material